Skip to Content
MilliporeSigma

MABC1145

Anti-FAP Antibody, clone 73.3

clone 73.3, from mouse

Synonym(s):

Fibroblast activation protein, Propyl endopeptidase FAP, Dipeptidyl peptidase FAP, Serine integral membrane protease, SIMP

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
73.3, monoclonal
Application:
FACS
Citations:
2
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

73.3, monoclonal

species reactivity

mouse

technique(s)

flow cytometry: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

mouse ... Fap(14089)

General description

87.94 kDa calculated.
Fibroblast activation protein (UniProt: ; also known as Propyl endopeptidase FAP, Dipeptidyl peptidase FAP, Serine integral membrane protease, SIMP) is encoded by the Fap gene (Gene ID: 14089) in murine species. FAP is a homodimeric type II transmembrane protein that physically attaches to cells and bulk of the protein is exposed to extracellular space. It is a gelatinase of the serine protease family that is selectively expressed by tumor associated fibroblasts and pericytes in a majority of human epithelial cancers, granulation tissue of healing wounds, and malignant cells of bone and soft tissue sarcomas. It is not expressed in most normal tissues, but is expressed during embryonic development. It is present as a homodimer and homodimerization is required for activity of both plasma membrane and soluble forms. It can also form heterodimer with DPP4. Both plasma membrane and soluble forms exhibit post-proline cleaving endopeptidase activity, with a marked preference for Ala/Ser-Gly-Pro-Ser/Asn/Ala consensus sequences. Its gelatinase activity is inhibited by several serine-protease inhibitors, such as PMSF, AEBSF, and diisopropyl fluorophosphate (DFP). FAP is involved in the control of fibroblast growth or epithelial-mesenchymal interactions during development, tissue repair, and epithelial carcinogenesis. It is also involved in the cell invasiveness towards the ECM in malignant melanoma cancers. (Ref.: Kelly, T et al. (2012). Int. Rev. Cell Mol. Biol. 297:83-116; Santos, AM et al. (2009). J. Clin. Invest. 119(12):3613-3625).

Immunogen

FAP-expressing 3T3 cells.

Application

Detect fibroblast activation protein using this mouse monoclonal Anti-FAP Antibody, clone 73.3, Cat. No. MABC1145. It has been tested in Flow Cytometry.
Flow Cytometry Analysis: A representative lot detected FAP in Flow Cytometry applications (Wang, L.C., et. al. (2014). Cancer Immunol Res. 2(2):154-66).

Flow Cytometry Analysis: 1 µg/mL from a representative lot detected FAP in 3T3/FAP overexpressed cells, but not in 3T3 WT cells. (Courtesy of Leslie Hopper, University of Pennsylvania, USA).
Research Category
Apoptosis & Cancer

Biochem/physiol Actions

Clone 73.3 specifically detects fibroblast activation protein (FAP) in 3T3 cells overexpressing FAP.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Isotype testing: Identity Confirmation by Isotyping Test.


Isotyping Analysis: The identity of this monoclonal antibody is confirmed by isotyping test to be mouse IgG1 .

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library