Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABC1102

Sigma-Aldrich

Anti-DKK2 Antibody, clone 5F8

clone 5F8, from mouse

Synonym(s):

Dickkopf-related protein 2, Dickkopf-2, hDkk-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5F8, monoclonal

species reactivity

human

species reactivity (predicted by homology)

monkey (based on 100% sequence homology)

packaging

antibody small pack of 25 μL

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... DKK2(27123)

General description

Dickkopf-related protein 2 (UniProt: Q9UBU2; also known as Dickkopf-2, DKK2, Dkk-2, hDkk-2) is encoded by the DKK2 (also known as UNQ682/PRO1316) gene (Gene ID: 27123) in human. DKK2 is a member of the Dickkopf family that is expressed in heart, brain, skeletal muscle, and lung. It is synthesized with a signal peptide (aa 1-33) that is subsequently cleaved off in the mature form. DKK2 is reported to antagonize canonical Wnt signaling by inhibiting LRP5/6 interaction with Wnt and by forming a ternary complex with the transmembrane protein KREMEN that promotes internalization of LRP5/6. Its C-terminal cysteine-rich domain mediates its interaction with LRP5 and LRP6. It plays play an important role in vertebrate development, where it locally inhibits Wnt regulated processes such as antero-posterior axial patterning, limb development, somitogenesis and eye formation. DKK2 secreted by tumor cells is shown to act on cytotoxic lymphocytes and inhibit STAT5 signaling by blocking STAT5 nuclear localization via LRP5, independent of LRP6. Loss of DKK2 is reported to activate natural killer (NK) cells and CD8+ T cells in tumors that can delay tumor progression and enhance the effects of PD-1 blockade. (Ref.: Xiao, Q., et al. (2018). Nat. Med. 24(3): 262-270).

Specificity

Clone 5F8 specifically detects human Dickkopf-related protein 2. It targets an epitope with in 15 amino acids from the N-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 15 amino acids from the N-terminal region of human Dickkopf-2 protein.

Application

Anti-DKK2, clone 5F8, Cat. No. MABC1102, is a mouse monoclonal antibody that detects Dickkopf-related protein 2 and has been tested for use in ELISA, Inhibits Activity/Function, and Western Blotting.
Inhibits Activity/Function Analysis: A representative lot inhibited DKK2-mediated, but not DKK1-mediated, antagonism of Wnt activation. (Xiao, Q., et. al. (2018). Nat Med. 24(3):262-270).


ELISA Analysis: A representative lot detected DKK2 in ELISA applications (Xiao, Q., et. al. (2018). Nat Med. 24(3):262-270).
Research Category
Apoptosis & Cancer

Quality

Evaluated by Western Blotting in human heart tissue lysates.

Western Blotting Analysis: A 1:250 dilution of this antibody detected DKK2 in human heart tissue lysates.

Target description

~38 kDa observed; 28.45 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in PBS without azide.

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service