Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB8906F

Sigma-Aldrich

Anti-Measles Antibody, nucleoprotein, clone 83KKII, FITC-conjugated

clone 83KKII, Chemicon®, from mouse

Synonym(s):

Rubeola

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

FITC conjugate

antibody form

purified antibody

antibody product type

primary antibodies

clone

83KKII, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunofluorescence: suitable

isotype

IgG

shipped in

wet ice

Specificity

MAB8906F recognizes the measles nucleoprotein by capture EIA.

Immunogen

Epitope: nucleoprotein

Application

Anti-Measles Antibody, nucleoprotein, clone 83KKII, FITC-conjugated is an antibody against Measles for use in ELISA & IF.
Capture EIA

IFA

Final working dilutions must be determined by end user.
Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral

Physical form

Purified by Protein-A Sepharose. 1 mg/mL in PBS with 0.1% sodium azide.

Storage and Stability

Maintain at 2-8°C.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Sepharose is a trademark of Cytiva

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuqi Jing et al.
Cancer gene therapy, 27(12), 910-922 (2020-04-02)
The tumor stroma acts as a barrier that limits the efficacy of systemically administered oncolytic viruses (OV). We previously demonstrated that stromal-selective, retargeted oncolytic measles viruses (MVs) delay in vivo tumor progression. To further characterize the contribution of stromal targeting
Thomas Mourez et al.
Virology, 419(2), 117-125 (2011-09-06)
We generated a replicating chimeric measles virus in which the hemagglutinin and fusion surface glycoproteins were replaced with the gp160 envelope glycoprotein of simian immunodeficiency virus (SIVmac239). Based on a previously cloned live-attenuated Schwarz vaccine strain of measles virus (MV)
Histologic and molecular correlates of fatal measles infection in children.
Jose Antonio Plaza, Gerard J Nuovo
Diagnostic molecular pathology : the American journal of surgical pathology, part B null
Brigitta M Laksono et al.
Journal of virology, 92(8) (2018-02-14)
Measles is characterized by a transient immune suppression, leading to an increased risk of opportunistic infections. Measles virus (MV) infection of immune cells is mediated by the cellular receptor CD150, expressed by subsets of lymphocytes, dendritic cells, macrophages, and thymocytes.
Alice Meignié et al.
Molecular & cellular proteomics : MCP, 20, 100049-100049 (2021-01-31)
Viruses manipulate the central machineries of host cells to their advantage. They prevent host cell antiviral responses to create a favorable environment for their survival and propagation. Measles virus (MV) encodes two nonstructural proteins MV-V and MV-C known to counteract

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service