Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB8690

Sigma-Aldrich

Anti-Hepatitis C Virus Antibody, aa 70-90, Core

Chemicon®, from mouse

Synonym(s):

HCV

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunofluorescence: suitable
immunohistochemistry: suitable (paraffin)

isotype

IgG1

shipped in

wet ice

Specificity

Specific to HCV core antigen

Immunogen

Epitope: a.a. 70-90
Recombinant protein to core antigen

Application

Detect Hepatitis C Virus using this Anti-Hepatitis C Virus Antibody, aa 70-90, Core validated for use in ELISA, IF, IH(P).
ELISA

Immunohistochemistry (Parrafin embedded)

Immunofluorescence

Final dilutions must be determined by end user

Physical form

Format: Purified
Purified immunoglobulin. Liquid 0.02MPB, 0.25M NaCl, 0.1% sodium azide

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service