Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB5264

Sigma-Aldrich

Anti-Nerve Growth Factor Receptor (p75) Antibody, clone 8211

clone 8211, Chemicon®, from mouse

Synonym(s):

NGF-Receptor p75, Low Affinity NGF Receptor, NGFR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

8211, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NTF3(4908)

General description

The low affinity NGFR (Nerve growth factor receptor) is a 75kDa membrane-spanning glycoprotein lacking intrinsic tyrosine kinase activity. p75NGFR interacts with TrkA, the high affinity NGF receptor and potentiates TrkA signaling at low NGF concentrations. The p75 receptor binds nerve growth factor, brain-derived neurotrophic factor, neurotrophin-3 and neurotrophin-4 with varying specificities. The p75NGFR plays an important role in neurotrophic factor signaling and has been shown to modulate the susceptibility of selective cellular populations to programmed cell death. It is expressed on many neuronal cells types including many embryonic forms and the receptor can be used to isolate neuronal progenitor cells.

Specificity

Reacts with NGF-receptors in human system.

Immunogen

Membrane fragments of the melanoma cell line WM-9 (Herlyn et al., 1983).

Application

Anti-Nerve Growth Factor Receptor (p75) Antibody, clone 8211 is an antibody against Nerve Growth Factor Receptor (p75) for use in IC, IH, IH(P) & WB.
Immunohistochemistry: 5-10 μg/mL

Immunocytochemistry: 5-10 μg/mL

Immunohistochemistry on paraffin sections: Cattoretti, Blood 81:1726-1738, 1993. http://www.bloodjournal.org/cgi/reprint/81/7/1726.pdf

Western Blot: 1ug/mL

Immunoabsorptive purification of NGF-receptors

Optimal working dilutions must be determined by end user.

Target description

75 kDa

Linkage

Replaces: 04-1111

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gaetana Minnone et al.
RMD open, 3(2), e000441-e000441 (2017-09-29)
To identify the role of mature nerve growth factor (mNGF), its immature form proNGF and their receptors in arthritis inflammation. Real-time PCR, western blot and ELISA were performed to evaluate NGF, proNGF, their receptor and cytokine expression in synovial tissue
Darren Carwardine et al.
Journal of the neurological sciences, 367, 311-318 (2016-07-18)
A multitude of factors must be overcome following spinal cord injury (SCI) in order to achieve clinical improvement in patients. It is thought that by combining promising therapies these diverse factors could be combatted with the aim of producing an
Mary-Louise Rogers et al.
Journal of neuroimmunology, 226(1-2), 93-103 (2010-06-16)
The common neurotrophin receptor P75NTR, its co-receptor sortilin and ligand proNGF, have not previously been investigated in Natural Killer (NK) cell function. We found freshly isolated NK cells express sortilin but not significant amounts of P75NTR unless exposed to interleukin-12
Daisuke Ito et al.
PloS one, 14(3), e0213252-e0213252 (2019-03-07)
Olfactory ensheathing cells are thought to support regeneration and remyelination of damaged axons when transplanted into spinal cord injuries. Following transplantation, improved locomotion has been detected in many laboratory models and in dogs with naturally-occurring spinal cord injury; safety trials
Darren Carwardine et al.
PloS one, 12(12), e0188967-e0188967 (2017-12-12)
Olfactory ensheathing cell (OEC) transplantation is a promising strategy for treating spinal cord injury (SCI), as has been demonstrated in experimental SCI models and naturally occurring SCI in dogs. However, the presence of chondroitin sulphate proteoglycans within the extracellular matrix

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service