Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB4703

Sigma-Aldrich

Anti-Caspase 3 Antibody, large subunit & proform, clone 4-1-18

clone 4-1-18, Chemicon®, from mouse

Synonym(s):

CPP32

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4-1-18, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CASP3(836)

Specificity

Recognizes the full length 32 kDa caspase-3 protein. Highly specific to caspase-3 and shows no cross-reaction with other caspase family members. Caspase-3 is ubiquitously expressed and like other caspases is synthesized as an inactive proenzyme. Upon activation, caspase-3 is cleaved generating two smaller subunits of 17 kDa and 12 kDa. Caspase-3 is one of the most extensively studied enzyme in apoptosis. Activation of caspase-3 occurs in response to a variety of apoptotic inducers.

Immunogen

Epitope: large subunit & proform
Recombinant full-length human caspase-3 protein

Application

Anti-Caspase 3 Antibody, large subunit & proform, clone 4-1-18 detects level of Caspase 3 & has been published & validated for use in WB, IH(P).
Research Category
Apoptosis & Cancer

Metabolism
Research Sub Category
Caspases

Enzymes & Biochemistry
Western blot: 1:1000. The antibody will detect a band at 32 kDa corresponding to the inactive unprocessed form of caspase 3 as well as a 17-22 kDa band corresponding to the large subunit of active caspase 3.

Immunohistochemistry (frozen and paraffin): 1:500.



Optimal working dilutions must be determined by end user.

Linkage

Replaces: 04-439

Physical form

Format: Purified
Purified immunoglobulin by protein A. Liquid in PBS, pH 7.6, with 0.1% sodium azide.

Storage and Stability

Maintain refrigerated at 2-8°C in undiluted aliquots for up to 12 months.

Analysis Note

Control
POSITIVE CONTROL: 293 (human embryonic kidney) cell lysates

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elmira R Mordakhanova et al.
International journal of molecular sciences, 21(7) (2020-04-11)
Heparin-induced thrombocytopenia (HIT) is an adverse drug reaction characterized by thrombocytopenia and a high risk for venous or arterial thrombosis. HIT is caused by antibodies that recognize complexes of platelet factor 4 and heparin. The pathogenic mechanisms of this condition
Tatiana A Nevzorova et al.
Cell death discovery, 5, 106-106 (2019-07-03)
Heparin-induced thrombocytopenia (HIT) is a complication of heparin therapy sometimes associated with thrombosis. The hallmark of HIT is antibodies to the heparin/platelet factor 4 (PF4) complex that cause thrombocytopenia and thrombosis through platelet activation. Despite the clinical importance, the molecular
Yan Sun et al.
Theranostics, 8(7), 2044-2060 (2018-03-21)
Rationale: Cardenolides have potential as anticancer drugs. 3'-epi-12β-hydroxyfroside (HyFS) is a new cardenolide structure isolated by our research group, but its molecular mechanisms remain poorly understood. This study investigates the relationship between its antitumor activities and autophagy in lung cancer
Oleg V Kim et al.
Haematologica, 104(9), 1866-1878 (2019-02-23)
Platelets play a key role in the formation of hemostatic clots and obstructive thrombi as well as in other biological processes. In response to physiological stimulants, including thrombin, platelets change shape, express adhesive molecules, aggregate, and secrete bioactive substances, but
Anna Chiarini et al.
International journal of molecular medicine, 36(3), 845-856 (2015-07-24)
Protein kinase (PK)Cζ signaling at various subcellular levels affects cell survival, differentiation, growth and/or apoptosis. However, the mechanisms modulating PKCζ activity at the nuclear membrane (NM) are not yet fully understood. Previously, we demonstrated that PKCζ interacts with the B‑cell

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service