Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB4601

Sigma-Aldrich

Anti-Bax Antibody, NT, aa3-16 hBax protein, clone 2D2

clone 2D2, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2D2, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... BAX(581)

General description

The previously assigned protein identifier Q07814 has been merged into Q07812. Full details can be found on the UniProt database.

Specificity

Recognizes a 21 kD protein. Does not cross-react with Bcl-2a or Bcl-X. In contrast to Bcl-2a and Bcl-X, overexpression of Bax protein accelerates apoptosis.

Immunogen

Epitope: N-terminus, aa3-16 hBax protein
Synthetic peptide corresponding to aa 3-16 (GSGEQPRGGGPTSS) of human bax. This amino acid sequence is not shared by mouse or rat Bax.

Application

Anti-Bax Antibody, N-terminus, aa3-16 hBax protein, clone 2D2 is an antibody against Bax for use in IF, WB, IH(P).
Research Category
Apoptosis & Cancer
Research Sub Category
BCL2 & Inhibition
Western blot: 1 μg/ml for 2 hours at RT (1:200 dilution)

Immunofluorescence

Immunohistochemistry (frozen and paraffin): 2-4 μg/ml for 30 minutes at RT

(Staining of formalin-fixed tissues requires boiling tissue sections in 10 mM citrate buffer, pH 6.0, for 10-20 min. followed by cooling for 20 min. at RT)

Optimal working dilutions must be determined by end user.

Linkage

Replaces: 04-434

Physical form

Format: Purified
Liquid in 10 mM PBS, pH 7.4, with 0.2% BSA and 15 mM sodium azide.

Storage and Stability

Maintain refrigerated at 2-8°C in undiluted aliquots for up to 12 months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hao Fang et al.
Neuroreport, 32(7), 577-582 (2021-04-15)
This study was to detect the protective effects of Houttuynia cordata extract on the damage induced by propofol in hippocampal neuron of rats. Propofol-induced neuron injury model and H. cordata extract administration were conducted. Immunofluorescence and immunoblot were conducted for
Opa1-mediated cristae opening is Bax/Bak and BH3 dependent, required for apoptosis, and independent of Bak oligomerization.
Yamaguchi, R; Lartigue, L; Perkins, G; Scott, RT; Dixit, A; Kushnareva, Y; Kuwana et al.
Molecular Cell null
Suzanne Gaudet et al.
PLoS computational biology, 8(4), e1002482-e1002482 (2012-05-10)
Stochastic fluctuations in gene expression give rise to cell-to-cell variability in protein levels which can potentially cause variability in cellular phenotype. For TRAIL (TNF-related apoptosis-inducing ligand) variability manifests itself as dramatic differences in the time between ligand exposure and the
Maotao Du et al.
Annals of translational medicine, 9(23), 1722-1722 (2022-01-25)
Aloe-emodin is reported as a potential cancer therapeutic agent due to its inhibition of the proliferation, migration, and invasion of cancer cells. This study aimed to confirm the effects of aloe-emodin on the progression of melanoma and identify the underlying
Marc Payton et al.
Molecular cancer therapeutics, 17(12), 2575-2585 (2018-09-30)
Aurora kinase A and B have essential and non-overlapping roles in mitosis, with elevated expression in a subset of human cancers, including acute myeloid leukemia (AML). In this study, pan-aurora kinase inhibitor (AKI) AMG 900 distinguishes itself as an anti-leukemic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service