Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

MAB377B

Sigma-Aldrich

Anti-NeuN Antibody, clone A60, biotin conjugated

clone A60, Chemicon®, from mouse

Synonym(s):

Neuron-Specific Nuclear Protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

biotin conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

A60, monoclonal

species reactivity

rat, mouse

species reactivity (predicted by homology)

ferret, salamander, human, chicken

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG1

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

NeuN antibody (NEUronal Nuclei; clone A60) specifically recognizes the DNA-binding, neuron-specific protein NeuN, which is present in most CNS and PNS neuronal cell types of all vertebrates tested. NeuN protein distributions are apparently restricted to neuronal nuclei, perikarya and some proximal neuronal processes in both fetal and adult brain although, some neurons fail to be recognized by NeuN at all ages: INL retinal cells, Cajal-Retzius cells, Purkinje cells, inferior olivary and dentate nucleus neurons, and sympathetic ganglion cells are examples (Mullen et al., 1992; Wolf et al., 1996). Immunohistochemically detectable NeuN protein first appears at developmental timepoints that correspond with the withdrawal of the neuron from the cell cycle and/or with the initiation of terminal differentiation of the neuron (Mullen et al., 1992). Immunoreactivity appears around E9.5 in the mouse neural tube and is extensive throughout the developing nervous system by E12.5. Strong nuclear staining suggests a nuclear regulatory protein function; however, no evidence currently exists as to whether the NeuN protein antigen has a function in the distal cytoplasm or whether it is merely synthesized there before being transported back into the nucleus. No difference between protein isolated from purified nuclei and whole brain extract on immunoblots has been found (Mullen et al., 1992).

Specificity

Vertebrate neuron-specific nuclear protein called NeuN (Neuronal Nuclei). MAB377B reacts with most neuronal cell types throughout the nervous system of mice including cerebellum, cerebral cortex, hippocampus, thalamus, spinal cord and neurons in the peripheral nervous system including dorsal root ganglia, sympathetic chain ganglia and enteric ganglia. The immunohistochemical staining is primarily in the nucleus of the neurons with lighter staining in the cytoplasm. The few cell types not reactive with MAB377B include Purkinje, mitral and photoreceptor cells. Developmentally, immunoreactivity is first observed shortly after neurons have become postmitotic, no staining has been observed in proliferative zones. The antibody is an excellent marker for neurons in primary cultures and in retinoic acid-stimulated P19 cells. It is also useful for identifying neurons in transplants.

Immunogen

Purified cell nuclei from mouse brain

Application

Immunocytochemistry Analysis:
1:10-1:500 dilution of a previous lot was used. Neurons in culture should be permeablized with 0.1% triton X-100. All primary antibody dilutions should be performed with simple solutions containing only buffer and primary antibody without excess protein blocks or detergents.

For dual labeling studies using mouse monoclonals, antibody incubations should be sequential with MAB377B last. First mouse monoclonal antibody should be first detected with anti-mouse secondary prior to incubating with MAB377B. Excess anti-mouse IgG may be blocked by incubating with 1% mouse serum prior to MAB377B incubation. Detection of biotinylated NeuN monoclonal is via streptavidin. In some cases in may be necessary to pretreat the tissue with avidin to block excess biotin prior to immunohistochemisty (Wood and Warnke, 1981).

Immunohistochemistry:
1:200-1:2,000. The antibody works best on polyester wax embedded tissue but also works on paraffin embedded tissue at a lower working dilution. The antibody works well with formaldehyde-based fixatives. Citric acid and microwave pretreatment has been used successfully (Sarnat, 1998).

Western Blotting Analysis:
A previous lot of this antibody was used on western blot. Recognizes 2-3 bands in the 46-48 kDa range and possibly another band at approximately 66 kDa.

Optimal working dilutions must be determined by end user.
Research Category
Neuroscience
Research Sub Category
Neuronal & Glial Markers
This Anti-NeuN Antibody, clone A60, biotin conjugated is validated for use in IC, IH, IH(P), WB for the detection of NeuN.

Quality

Routinely evaluated by immunohistochemistry on brain tissue.

Immunohistochemistry(paraffin) Analysis:
NeuN (cat. # MAB377B) staining pattern/morphology in rat cerebellum. Tissue pretreated with Citrate, pH 6.0. This lot of antibody was diluted to 1:100, using IHC-Select Detection with HRP-DAB. Immunoreactivity is seen as nuclear staining in the neurons in the granular layer. Note that there is no signal detected in the nucleus of Purkinje cells.
Optimal Staining With Citrate Buffer, pH 6.0, Epitope Retrieval: Rat Cerebellum

Target description

46-48 kDa

Physical form

Protein A purified
Purified mouse monoclonal IgG1 in buffer containing 0.01 M PBS pH 7.1, 0.1% sodium azide with 15 mg/mL BSA as a stabilizer.

Storage and Stability

Stable for 1 year at 2-8ºC from date of receipt.

Analysis Note

Control
Brain tissue, most neuronal cell types throughout the adult nervous system

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

c-Fos expression during temporal order judgment in mice.
Wada, M; Higo, N; Moizumi, S; Kitazawa, S
Testing null
Chronic IL-1?-mediated neuroinflammation mitigates amyloid pathology in a mouse model of Alzheimer's disease without inducing overt neurodegeneration.
Matousek, SB; Ghosh, S; Shaftel, SS; Kyrkanides, S; Olschowka, JA; O'Banion, MK
Journal of Neuroimmune Pharmacology null
Motor unit abnormalities in Dystonia musculorum mice.
De Repentigny, Y; Ferrier, A; Ryan, SD; Sato, T; Kothary, R
Testing null
Jean-Luc Boulland et al.
PloS one, 8(8), e71701-e71701 (2013-08-31)
Despite limited regeneration capacity, partial injuries to the adult mammalian spinal cord can elicit variable degrees of functional recovery, mediated at least in part by reorganization of neuronal circuitry. Underlying mechanisms are believed to include synaptic plasticity and collateral sprouting
Jing Chen-Roetling et al.
Neurobiology of disease, 102, 140-146 (2017-03-23)
Pharmacotherapies that increase CNS expression of heme oxygenase-1 (HO-1) and other antioxidant proteins have improved outcome in experimental models of spontaneous intracerebral hemorrhage (ICH). In order to more specifically investigate the relationship between HO-1 and ICH outcome, mice expressing human

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service