Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB3236

Sigma-Aldrich

Anti-Cytokeratin 18 Antibody, clone RCK106

clone RCK106, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
RCK106, monoclonal
application:
FACS
ICC
IHC
WB
species reactivity:
human
technique(s):
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable
citations:
2

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

RCK106, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... KRT18(3875)

General description

Cytokeratins are a subfamily of intermediate filament proteins and are characterized by a remarkable biochemical diversity, represented in epithelial tissues by at least 20 different polypeptides. They range in molecular weight from between 40 kDa and 68 kDa and isoelectric pH between 4.9 - 7.8, The individual cytokeratin polypeptides are designated 1-20. The various epithelia in the human body usually express cytokeratins which are not only characteristic of the type of epithelium, but also related to the degree of maturation or differentiation within an epithelium. Cytokeratin subtype expression patterns are used to an increasing extent in the distinction of different types of epithelial malignancies.

Specificity

Cytokeratin 18. MAB3236 reacts exclusively with human glandular epithelial cells of the digestive, respiratory, and urogenital tracts, endocrine and exocrine cells and mesothelial cells as well as adenocarcinomas originating from them.

SPECIES REACTIVITY:

Human only. Does not react with other species.

Immunogen

Cytoskeletal preparation of human bladder carcinoma cell line T24.

Application

Anti-Cytokeratin 18 Antibody, clone RCK106 detects level of Cytokeratin 18 & has been published & validated for use in FC, WB, IC, IH(P).
Research Category
Cell Structure
Research Sub Category
Cytokeratins
Western blot

Immunohistochemistry on frozen and paraffin embedded tissue sections.

Immunocytochemistry

Flow cytometry

Optimal working dilutions must be determined by the end user.

Linkage

Replaces: 04-586

Physical form

Format: Purified
Liquid in buffer with 0.1% sodium azide.

Storage and Stability

Maintain at -20°C in undiluted aliquots up to 6 months. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pukkavadee Netsirisawan et al.
International journal of oncology, 56(6), 1387-1404 (2020-04-03)
Breast cancer is the most common type of cancer and leading cause of cancer‑associated mortality in women worldwide. O‑linked N‑acetyl glucosaminylation (O‑GlcNAcylation) is a dynamic post‑translational modification of nuclear, cytoplasmic and mitochondrial proteins. Mounting evidence suggests that abnormal O‑GlcNAcylation status
Weixuan Wang et al.
Cell chemical biology, 25(3), 255-261 (2018-01-09)
Nicotinamide adenine dinucleotide (NAD) levels decrease with aging as a result of aging-associated CD38 upregulation. Here, we established a cell model with decreased cellular NAD levels by overexpressing CD38 or treating cells with FK866, an inhibitor of nicotinamide phosphoribosyltransferase. We

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service