Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB317

Sigma-Aldrich

Anti-Calcitonin Gene Related Peptide Antibody, α hCGRP a.a. 28-37, clone CD8

clone CD8, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

CD8, monoclonal

species reactivity

guinea pig, human

should not react with

rat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CALCA(796)

Specificity

Highly specific for CGRP and does not cross-react with other amino acids.

Immunogen

A terminal fragment of hCGRP coupled to BSA with glutaraldehyde.
Epitope: alpha hCGRP a.a. 28-37

Application

Anti-Calcitonin Gene Related Peptide Antibody, α hCGRP a.a. 28-37, clone CD8 is an antibody against Calcitonin Gene Related Peptide for use in ELISA, IH.
Immunohistochemistry at 1:200-1:400. Antibody not reactive on formalin fixed tissues. 4% PFA frozen tissues preferred. Immersion (2 hours) or perfused (pH7.4); Blocking in PBS with 3% BSA or NGS containing 0.1% triton X-100 is advised. Primary antibody should be diluted only in PBS with no detergent or blocking compounds.

ELISA: against BSA-conjugated alpha CGRP peptide.



Optimal working dilutions to be determined by end user.
Research Category
Neuroscience
Research Sub Category
Hormones & Receptors

Physical form

Format: Purified

Storage and Stability

Maintain refrigerated at 2-8°C for up to 12 months.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katarzyna Palus et al.
International journal of molecular sciences, 21(1) (2019-12-22)
The digestive tract, especially the small intestine, is one of the main routes of acrylamide absorption and is therefore highly exposed to the toxic effect of acrylamide contained in food. The aim of this experiment was to elucidate the effect
Do-Hee Kim et al.
Scientific reports, 7(1), 15214-15214 (2017-11-11)
Acupuncture, a traditional medical procedure practised for over 2000 years in Asia, stimulates specific but poorly defined sites called acupoints. To date, no unique anatomical acupoint structures have been found. However, noxious sensory signals from visceral organs produce hypersensitive spots
Glyphosate affects the neurochemical phenotype of the intramural neurons in the duodenum in the pig.
Palus, et al.
Neurogastroenterology and Motility, 35, e14507-e14507 (2023)
K J Dougherty et al.
Neuroscience, 163(3), 909-919 (2009-06-30)
The synthesis enzyme glutamic acid decarboxylase (GAD65 or GAD67) identifies neurons as GABAergic. Recent studies have characterized the physiological properties of spinal cord GABAergic interneurons using lines of GAD67-green fluorescent protein (GFP) transgenic mice. A more complete characterization of their
Guo-Du Wang et al.
American journal of physiology. Gastrointestinal and liver physiology, 307(7), G719-G731 (2014-08-26)
Mast cells express the substance P (SP) neurokinin 1 receptor and the calcitonin gene-related peptide (CGRP) receptor in guinea pig and human small intestine. Enzyme-linked immunoassay showed that activation of intramural afferents by antidromic electrical stimulation or by capsaicin released

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service