Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB2616

Sigma-Aldrich

Anti-Kindlin-1 Antibody, clone 4A5.14

clone 4A5.14, from mouse

Synonym(s):

Kindlin syndrome protein, UNC-112 related protein 1, Unc-112-related protein 1, chromosome 20 open reading frame 42, kindlin 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4A5.14, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FERMT1(55612)

General description

The Kindlin family is a group of focal adhesion proteins that have recently become a point of interest because of their ability to bind and activate integrins. Abnormalities in this ability have linked Kindlin to several diseases, both inherited and acquired, such as Kindler syndrome, cancer, and leukocyte adhesion deficiency. Specifically associated with Kindler syndrome, is the isoform Kindlin-1 which plays a key role in adhering the actin cytoskeleton to the extracellular matrix of basal keratinocytes. Kindlin-1 may also be involved in tumor growth by modulating TGF-beta 1 signaling (Shi, 2007). It is most widely expressed in the brain, kidney, colon, placenta, skeletal muscle, adrenal gland, and prostate.

Specificity

The antibody recognizes Kindlin-1.

Immunogen

Epitope: Unknown
GST-tagged recombinant protein corresponding to human Kindlin-1.

Application

Anti-Kindlin-1 Antibody, clone 4A5.14 is an antibody against Kindlin-1 for use in WB, IC, IH.
Immunocytochemistry Analysis: A previous lot was used by an independent laboratory in IC. (Papachristou, D.J., et al. (2007). Histopathology. 51(4):499–508.)

Immunohistochemistry Analysis: A previous lot was used by an independent laboratory in IH. (Papachristou, D.J., et al. (2007). Histopathology. 51(4):499–508.)
Research Category
Cell Structure
Research Sub Category
Cytoskeleton

Quality

Evaluated by Western Blot in HeLa cell lysate.

Western Blot Analysis: 0.5 µg/ml of this antibody detected Kindlin-1 in 10 µg of HeLa cell lysate.

Target description

~ 70 kDa

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Baisong Zhao et al.
The journal of headache and pain, 18(1), 1-1 (2017-01-07)
Hyperbaric oxygen (HBO) therapy is proven to attenuate neuropathic pain in rodents. The goal of the present study was to determine the potential involvement of the Kindlin-1/Wnt-10a signaling pathway during astrocyte activation and inflammation in a rodent model of neuropathic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service