Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

MAB1614

Sigma-Aldrich

Anti-Kinesin Antibody, heavy chain, a.a.420-445, clone H2

clone H2, Chemicon®, from mouse

Synonym(s):

Anti-HEL-S-61, Anti-KINH, Anti-KNS, Anti-KNS1, Anti-UKHC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

H2, monoclonal

species reactivity

sea urchin, rat, chicken, squid, human, mouse

species reactivity (predicted by homology)

mammals

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
radioimmunoassay: suitable
western blot: suitable

isotype

IgG2b

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... KIF5B(3799)

General description

Kinesins are molecular motors used for intracellular transport and trafficking. They move along microtubule cables powered by the hydrolysis of ATP, thus kinesins are ATPases. The active movement of kinesins supports several cellular functions including mitosis, meiosis and transport of cargo such as axonal transport.

Specificity

Reacts with the heavy chain of kinesin near the N-terminus.

Immunogen

Bovine brain kinesin.
Epitope: a.a.420-445

Application

Detect Kinesin using this Anti-Kinesin Antibody, heavy chain, a.a.420-445, clone H2 validated for use in ELISA, IC, IH, IP, FUNC, RIA & WB.
Immunohistochemistry:
A previous lot was used on paraformaldehyde fixed tissue. Also works for EM.

Immunocytochemistry:
A previous lot of this antibody was used in Immunocytochemistry.

Immunoprecipitation:
A previous lot of this antibody was used in Immunoprecipitation.

EIA/RIA:
A previous lot of this antibody was used in RIA.

Immunoblotting: recognizes 124kDa KHC and its proteolytic fragments.

Inhibition of kinesin mediated transport {Brady, et al PNAS 87(3):1061-1065, 1990.

Optimal working dilutions must be determined by the end user.
Research Category
Cell Structure
Research Sub Category
Cytoskeleton

Quality

Routinely evaluated by Western Blot on Mouse Brain lysates.

Western Blot Analysis:
1:500 dilution of this lot detected KINESIN (HEAVY CHAIN) on 10 μg of Mouse Brain lysates.

Target description

124 kDa

Physical form

Format: Purified
Protein A purified
Purified mouse monoclonal IgG2b in buffer containing 0.02 M Phosphate buffer, 0.25 M NaCl with 0.1% sodium azide.

Storage and Stability

Stable for 6 months at 2-8°C in undiluted aliquots from date of receipt.

Analysis Note

Control
Brain tissue

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Opposing microtubule motors drive robust nuclear dynamics in developing muscle cells.
Wilson, MH; Holzbaur, EL
Journal of Cell Science null
T Lynne Blasius et al.
PloS one, 8(9), e76081-e76081 (2013-10-08)
Kinesin motors drive the long-distance anterograde transport of cellular components along microtubule tracks. Kinesin-dependent transport plays a critical role in neurogenesis and neuronal function due to the large distance separating the soma and nerve terminal. The fate of kinesin motors
A non-mammalian type opsin 5 functions dually in the photoreceptive and non-photoreceptive organs of birds.
Ohuchi, H; Yamashita, T; Tomonari, S; Fujita-Yanagibayashi, S; Sakai, K; Noji, S; Shichida, Y
Testing null
Muhammad Saad Yousuf et al.
eNeuro, 6(2) (2019-04-09)
Multiple sclerosis (MS) is an autoimmune, demyelinating disease of the central nervous system. Patients with MS typically present with visual, motor, and sensory deficits. However, an additional complication of MS in large subset of patients is neuropathic pain. To study
Tadashi Nakaya et al.
Scientific reports, 8(1), 15575-15575 (2018-10-24)
Amyotrophic Lateral Sclerosis (ALS) is a lethal neurodegenerative disorder that primarily affects motor neurons. Dominant mutations in the RNA binding protein Fused in Sarcoma (FUS) have been identified as causative factors of ALS. Mutation, R495X, results in a premature stop

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service