Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MAB1273A4

Sigma-Aldrich

Anti-Mitochondria Antibody, clone 113-1, Alexa Fluor 488 conjugate

clone 113-1, from mouse, ALEXA FLUOR 488

Synonym(s):

Human Mitochondria

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

ALEXA FLUOR 488

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

113-1, monoclonal

species reactivity

human (mitochondria protein)

should not react with

mouse (mitochondria protein), rat (mitochondria protein)

technique(s)

immunocytochemistry: suitable

isotype

IgG1

shipped in

wet ice

target post-translational modification

unmodified

General description

This antibody recognizes a 60 kDa nonglycosylated protein component of mitochondria found in human cells. It can be used to stain mitochondria in cell, tissue or biochemical preparations and can be used as a marker of the mitochondria in subcellular fractions. MAB1273A4 recognizes a 65 kD protein by immunoprecipitation and produces the stringy, "spaghetti-like", staining patten in the cytoplasm of human cells.

Application

Detect Mitochondrial Marker using this mouse monoclonal antibody, Anti-Mitochondria Antibody, clone 113-1 & Alexa Fluor 488 conjugate validated for use in ICC.
Evaluated by Immunocytochemistry in primary mouse embryonic fibroblasts (PMEFs).
Immunocytochemsitry Analysis: A 1:50 dilution of this antibody did not detect mitrochondria in primary mouse embryonic fibroblasts (PMEFs).

Quality

Evaluated by Immunocytochemistry in human adipose mesenchymal stem cells (SCC038).
Immunocytochemsitry Analysis: A 1:50 dilution of this antibody detected mitrochondria in human adipose mesenchymal stem cells (SCC038).

Target description

60 kDa calculated

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna C Krieger et al.
Cancers, 15(10) (2023-06-22)
Cardiolipin (CL) is a mitochondrial lipid with diverse roles in cellular respiration, signaling, and organelle membrane structure. CL content and composition are essential for proper mitochondrial function. Deranged mitochondrial energy production and signaling are key components of glial cell cancers
Markus Pasztorek et al.
Frontiers in bioengineering and biotechnology, 7, 338-338 (2019-12-06)
The mechanobiological behavior of mesenchymal stem cells (MSCs) in two- (2D) or three-dimensional (3D) cultures relies on the formation of actin filaments which occur as stress fibers and depends on mitochondrial dynamics involving vimentin intermediate filaments. Here we investigate whether
Hao Chen et al.
Research (Washington, D.C.), 2021, 9798580-9798580 (2021-07-13)
Stem cell therapy holds high promises in regenerative medicine. The major challenge of clinical translation is to precisely and quantitatively evaluate the in vivo cell distribution, migration, and engraftment, which cannot be easily achieved by current techniques. To address this
Jialing Zhang et al.
Cancer research, 76(22), 6588-6597 (2016-11-05)
Oncocytic tumors are characterized by an excessive eosinophilic, granular cytoplasm due to aberrant accumulation of mitochondria. Mutations in mitochondrial DNA occur in oncocytic thyroid tumors, but there is no information about their lipid composition, which might reveal candidate theranostic molecules.
Estelle Porret et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 182, 141-151 (2022-12-19)
Glioblastoma (GBM) is the most common and deadly form of primary brain tumor. Between 30 % and 60 % of GBM are characterized by overexpression of the Epidermal Growth Factor Receptor (EGFR). The anti-EGFR antibody Cetuximab (CTX) showed a favorable effect for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service