Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB1096

Sigma-Aldrich

Anti-Tumor Necrosis Factor-α Antibody, clone 195

clone 195, Chemicon®, from mouse

Synonym(s):

TNF-alpha

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

195, monoclonal

species reactivity

human

should not react with

pig, monkey, canine, mouse, rat, baboon

packaging

antibody small pack of 25 μg

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
neutralization: suitable
western blot: suitable

isotype

IgG3

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TNF(7124)

Specificity

The antibody reacts with human, natural and recombinant TNF-alpha. lt does not react with human TNF-beta (lymphotoxin). lt does not react with TNF-alpha from mouse, rat, canine, porcine, rhesus-monkey or baboon.

SPECIES REACTIVITIES:

Not mouse, rat, canine, porcine, baboon or rhesus-monkey.

Immunogen

Human, recombinant tumor necrosis factor-alpha produced in E. coli.

Application

Research Category
Signaling
Research Sub Category
Growth Factors & Receptors
This Anti-Tumor Necrosis Factor-α Antibody, clone 195 is validated for use in IC, IH, NEUT, WB for the detection of Tumor Necrosis Factor-α.
Western Blot

Immunocytochemistry

Immunohistochemistry

Neutralization: 50 ng of TNF-alpha antibody is required for the neutralization of 1 U TNF-alpha. The TNF-alpha-preparation which is used has a specific activity of 2 x 10(7) U/mg protein [cell lytic assay with L929 cells (mouse transformed fibroblasts) and/or WEHJ 164 cells (mouse fibrosarcoma cells)/actinomycin D].

Optimal working dilutions must be determined by end user.

Linkage

Replaces: 04-1114

Physical form

Format: Purified
Purified immunoglobulin. Liquid in 0.01M PBS pH 7.1, 0.15M NaCl containing no preservatives. Sterile filtered.

Storage and Stability

Maintain frozen at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles. Freeze only once.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Micaela Morgado et al.
Oncotarget, 7(12), 14871-14884 (2016-02-27)
Transmembrane mucins (TMs) are restricted to the apical surface of normal epithelia. In cancer, TMs not only are over-expressed, but also lose polarized distribution. MUC16/CA125 is a high molecular weight TM carrying the CA125 epitope, a well-known molecular marker for
Mei Liu et al.
International journal of oncology, 38(4), 1013-1022 (2011-01-29)
Many studies have demonstrated the overexpression and amplification of the miR-17-92 cluster in malignant human cancers, including B-cell lymphomas and lung cancers. The purpose of this study was to investigate for the first time, the expression of the miR-17-92 cluster
Emanuele Garzia et al.
Fertility and sterility, 100(1), 108-115 (2013-04-02)
To investigate the expression of prolactin (PRL), PRL-receptor (PRL-R), and the TH1 cytokines interleukin-2 (IL-2), tumor necrosis factor-α (TNF-α), and interferon-γ (IFN-γ) at the maternofetal interface. Case-control study. University hospital unit of gynecology and obstetrics and research laboratories. Women undergoing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service