Skip to Content
MilliporeSigma

MAB045

Anti-Fluorescein (FITC) Antibody, clone 5D6.2

clone 5D6.2, Chemicon®, from mouse

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Clone:
5D6.2, monoclonal
Technique(s):
ELISA: suitable
Application:
ELISA
Citations:
1
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5D6.2, monoclonal

species reactivity (predicted by homology)

all

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable

isotype

IgG2a

shipped in

wet ice

target post-translational modification

unmodified

Application

EIA. Optimal working dilutions must be determined by end user.
This Anti-Fluorescein (FITC) Antibody, clone 5D6.2 is validated for use in ELISA for the detection of Fluorescein (FITC).

Biochem/physiol Actions

Fluorescein (FITC)

Physical form

Format: Purified
Liquid in 0.02 M phosphate buffer, 0.25 M NaCl, pH 7.6, 0.1% sodium azide.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Replaces: CBL97

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Try our Product Selector Tool to narrow your options


wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

Storage Class

12 - Non Combustible Liquids



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
MAB04504053252505355