Skip to Content

IPAKKITEX

Milli-Q® Polishing Kit for Elix® Pure Water

greener alternative

Greener Alternative

For Milli-Q® IQ/EQ 7000 ultrapure water systems fed by Elix® pure water. This product replaces IPAKKITA1.

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41104212
NACRES:
JA.13
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

packaging

units of 1

manufacturer/tradename

Milli-Q®

storage condition

room temperature

greener alternative product characteristics

Waste Prevention
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

cartridge H

39.5 cm (15.5 in.)

input

feed water nature pure water (Elix® water)

compatibility

for use with Milli-Q® EQ 7000 (Feed Water from Elix®), for use with Milli-Q® IQ 7000 (Feed Water from Elix®)

greener alternative category

shipped in

ambient

General description

This purification kit contains IPAK Meta® and IPAK Quanta® polishing cartridges for Elix® water, designed to work as a pair to produce ultrapure (Type 1) water for sensitive laboratory applications.
The cartridges contain a combination of Jetpore® mixed-bed ion-exchange resin and innovative IQnano ion-exchange media to remove trace ions from purified water, as well as a high-grade synthetic activated carbon, which targets trace organic contaminants.
We continue to demonstrate our commitment to a sustainable future by exploring ways to improve existing products in our portfolio. We have reviewed our purification cartridges to identify key opportunities to minimize impact without compromising on the product performance. This product has benefited from an optimization of the resin mix, the packaging and the end-of-life management. Therefore, it is identified as a greener alternative product in the category reengineered. View the DfS scorecard for the new Milli-Q® polishing kit for Elix® water.

Application

  • For Milli-Q® IQ/EQ 7000 ultrapure water systems fed with Elix® feed water.
  • Removes trace ions and organic contaminants to deliver ultrapure (Type 1) water for sensitive applications.

Features and Benefits

  • e-Sure tags enable RFID connection with the system, for seamless consumable status monitoring, data management and full traceability.
  • Supplied with one Drain Cap per cartridge, to remove excess water and reduce the weight of exhausted cartridges at disposal.
  • 78% of the parts suppliers for IPAK Quanta® and IPAK Meta® cartridges included in this kit are ISO 14001 environmental management system-certified.

Legal Information

ELIX is a registered trademark of Merck KGaA, Darmstadt, Germany
IPAK META is a registered trademark of Merck KGaA, Darmstadt, Germany
IPAK QUANTA is a registered trademark of Merck KGaA, Darmstadt, Germany
IQNANO is a trademark of Merck KGaA, Darmstadt, Germany
JETPORE is a registered trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library