Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

IHCR1001-6

Sigma-Aldrich

Anti-Neuron-Specific Nuclear Protein Antibody, prediluted, clone A60

clone A60, Chemicon®, from mouse

Synonym(s):

NeuN

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

A60, monoclonal

species reactivity

pig, mouse, avian, salamander, rat, primate, human, vertebrates, chicken, ferret

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable (paraffin)

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... RBFOX3(146713)

General description

NeuN antibody (NEUronal Nuclei; clone A60) specifically recognizes the DNA-binding, neuron-specific protein NeuN, which is present in most CNS and PNS neuronal cell types of all vertebrates tested. NeuN protein distributions are apparently restricted to neuronal nuclei, perikarya and some proximal neuronal processes in both fetal and adult brain although, some neurons fail to be recognized by NeuN at all ages: INL retinal cells, Cajal-Retzius cells, Purkinje cells, inferior olivary and dentate nucleus neurons, and sympathetic ganglion cells are examples (Mullen et al., 1992; Wolf et al., 1996). Immunohistochemically detectable NeuN protein first appears at developmental timepoints that correspond with the withdrawal of the neuron from the cell cycle and/or with the initiation of terminal differentiation of the neuron (Mullen et al., 1992). Immunoreactivity appears around E9.5 in the mouse neural tube and is extensive throughout the developing nervous system by E12.5. Strong nuclear staining suggests a nuclear regulatory protein function; however, no evidence currently exists as to whether the NeuN protein antigen has a function in the distal cytoplasm or whether it is merely synthesized there before being transported back into the nucleus. No difference between protein isolated from purified nuclei and whole brain extract on immunoblots has been found (Mullen et al., 1992).

Specificity

neuron-specific protein NeuN

Immunogen

Purified cell nuclei from mouse brain.

Application

Anti-Neuron-Specific Nuclear Protein Antibody, prediluted, clone A60 detects level of Neuron-Specific Nuclear Protein & has been published & validated for use in IH(P).
Antibody is prediluted and ready to use for Immunohistochemistry of formalin-fixed, paraffin-embedded tissues.

Pretreatment: Heat Induced Epitope Retrieval (HIER). Recommend Citrate Buffer, pH 6.0 (Cat. No. 21545). No signal was detected without Epitope retrieval.

Incubation: 30 minutes with IHC Select Detection Kits.

NeuN, has been prediluted for use as the primary antibody with Chemicon′s IHC Select Detection Kits and Protocols (Catalog Nos. DAB050, DET-HP1000, APR050, and DET-APR1000), but other supplier′s IHC detection systems may be used. For optimized protocol details, visit www.chemicon.com and select the protocols link under Cat. No.IHCR1001-6.
Research Category
Neuroscience
Research Sub Category
Neuronal & Glial Markers

Physical form

Format: Purified
Liquid diluted in PBS, pH 7.2 with stabilizers, 0.2% Tween 20, and 0.1% as preservative.

Storage and Stability

Maintain at 2-8°C. Refer to vial for expiration dating.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohammad Charehsaz et al.
Daru : journal of Faculty of Pharmacy, Tehran University of Medical Sciences, 24(1), 24-24 (2016-10-08)
Evaluations of silver in both nanoparticle (Ag-NPs) and ionic forms indicate some adverse effects on living organisms, but little is known about their potential for developmental toxicity. In this study, developmental toxicity of Ag-NPs (from 0.2 to 20 mg/kg/day) and ionic
Anna Brunn et al.
The American journal of pathology, 173(1), 93-105 (2008-06-07)
The pathogenesis of inflammatory autoimmune diseases of the peripheral nervous system, leading to demyelination and/or axonal damage, remains incompletely understood. In particular, it is controversial regarding the extent to which (i) autoimmune-mediated destruction of peripheral nerves results in secondary damage
Lateral Connectivity in the Olfactory Bulb is Sparse and Segregated.
Kim, DH; Phillips, ME; Chang, AY; Patel, HK; Nguyen, KT; Willhite, DC
Frontiers in Neural Circuits null
Melissa K McCoy et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 26(37), 9365-9375 (2006-09-15)
The mechanisms that trigger or contribute to loss of dopaminergic (DA) neurons in Parkinson's disease (PD) remain unclear and controversial. Elevated levels of tumor necrosis factor (TNF) in CSF and postmortem brains of PD patients and animal models of PD
Iris Schäffner et al.
Molecular psychiatry (2022-03-24)
The transcription factor FOXG1 serves pleiotropic functions in brain development ranging from the regulation of precursor proliferation to the control of cortical circuit formation. Loss-of-function mutations and duplications of FOXG1 are associated with neurodevelopmental disorders in humans illustrating the importance

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service