Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

IFX-CAL-K20

Millipore

xMAP INTELLIFLEX Cal Kit, 20 Uses, RUO

Synonym(s):

Calibration Kit, INTELLIFLEX Cal Kit, xMAP Calibration Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116010
NACRES:
NC.54

packaging

kit of 1

compatibility

for use with INTELLIFLEX RUO System and INTELLIFLEX-DRSE

storage temp.

2-8°C

General description

For importing target values from the xMAP® INTELLIFLEX Calibration and Performance Verification Kits

Application

Multiplexing

Components

  • Luminex Classification Calibrator Microspheres
  • MagPlex Classification Calibrator Microspheres
  • Luminex Reporter Calibrator Microspheres
  • Lot specific CD for importing target values
  • 28 strip wells

Legal Information

xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service