Skip to Content
MilliporeSigma

HATF07850

Immobilon® -NC Nitrocellulose Membrane

50 sheets, 7 cm x 8.4 cm, 0.45 µm pore size, Triton-free, nitrocellulose-based transfer membrane

Synonym(s):

Western blotting membrane, blotting membrane, nitrocellulose transfer membrane, transfer membrane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41105339
eCl@ss:
32031602
NACRES:
NB.24
Material:
mixed cellulose esters (MCE) membrane, nitrocellulose membrane, plain filter, white filter
Detection method:
colorimetric, fluorometric, radioactive
Pore size:
0.45 μm pore size
Compatibility:
for use with Amido black, for use with CPTS, for use with Colloidal gold, for use with India ink, for use with Ponceau¬S red
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Immobilon®-NC Transfer Membrane Mini Sheets, 50 sheets, 7 cm x 8.4 cm, 0.45 µm pore size, Triton-free, mixed cellulose esters transfer membrane

Quality Segment

material

mixed cellulose esters (MCE) membrane, nitrocellulose membrane, plain filter, white filter

feature

hydrophilic

manufacturer/tradename

Immobilon®

technique(s)

electrophoresis transfer: suitable, western blot: suitable

filter L × W

7 cm × 8.4 cm

pore size

0.45 μm pore size

capacity

259 μg/cm2 adsorption capacity (goat IgG)

compatibility

for use with Amido black, for use with CPTS, for use with Colloidal gold, for use with India ink, for use with Ponceau¬S red

detection method

colorimetric, fluorometric, radioactive

shipped in

ambient

General description

Immobilon®-NC (HATF) membrane is composed of a mixed cellulose esters matrix.

Application

Immobilon®-NC Transfer Membrane has been used in western blot analysis.

Legal Information

Immobilon is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?


Pictograms

Flame

Signal Word

Danger

Hazard Codes

Hazard Classifications

Flam. Sol. 1

Storage Class

4.1B - Flammable solid hazardous materials



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library