Skip to Content
MilliporeSigma
All Photos(2)

Documents

GTTP14250

Millipore

Polycarbonate Membrane Filter, 0.22 μm Pore Size

Isopore, filter diam. 142 mm, hydrophilic

Synonym(s):

Isopore Membrane Filter

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602
NACRES:
NB.24

material

plain filter
polycarbonate membrane
white filter

sterility

non-sterile

feature

hydrophilic

manufacturer/tradename

Isopore
Millipore

parameter

0.007 L/min-cm2-psi air flow rate
140 °C max. temp.
3.36 mL/min-cm2-psi water flow rate (maximum flow rate per 1.0 psi)
~16 mL/min-cm2 water flow rate at 10 psi (typical results)

filter diam.

142 mm

thickness

25 μm

gravimetric extractables

<1%

refractive index

n/D 1.6

matrix

Isopore

pore size

0.22 μm pore size
13.8 % porosity

bubble point

≥5.2 bar, air with water at 23 °C

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

The Isopore membrane is composed of a polycarbonate film, with a smooth, glass-like surface. This enables a clearer sample observation in optical or electron microscopy. It is useful in the analysis of airborne contaminants. Also, the Isopore membrane has a precise pore diameter and a consistent pore size. It may find applications in epifluorescent microscopy and chemotaxis assays.

Application

  • Chemotaxis
  • Bioassays
  • Cytology
  • Air monitoring
  • SEM analysis
  • Sterility testing
  • Microplastics analysis
  • Microplastics analysis grade water

Isopore Membrane Filter has been used:
  • for the separation of respirable particles from sanded composites
  • to filter particulate organic matter (POM) from seawater
  • to concentrate the prokaryotic biomass from ocean water sample

Features and Benefits

  • Particle retention on the surface for easy counting and analysis.
  • Isopore membranes display low background interference.
  • Rapid drying and minimal sample analysis time.
  • Translucent material.
  • Also available in a brown variety.

Caution

Possible reduction in flow rates may occur after autoclaving.

Legal Information

ISOPORE is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bacterial assemblages of the eastern Atlantic Ocean reveal both vertical and latitudinal biogeographic signatures
Friedline CJ, et al.,
Biogeosciences (Online), 9(6) (2012)
Lindsey Bishop et al.
ACS nano, 11(9), 8849-8863 (2017-08-02)
Pulmonary toxicity studies on carbon nanotubes focus primarily on as-produced materials and rarely are guided by a life cycle perspective or integration with exposure assessment. Understanding toxicity beyond the as-produced, or pure native material, is critical, due to modifications needed
Silvia Vidal-Melgosa et al.
Nature communications, 12(1), 1150-1150 (2021-02-21)
The formation of sinking particles in the ocean, which promote carbon sequestration into deeper water and sediments, involves algal polysaccharides acting as an adhesive, binding together molecules, cells and minerals. These as yet unidentified adhesive polysaccharides must resist degradation by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service