Skip to Content
MilliporeSigma

GSWP14250

MCE Membrane Filter, 0.22 μm Pore Size

MF-Millipore, filter diam. 142 mm, hydrophilic, white

Synonym(s):

MF-Millipore Membrane Filter, 0.22 µm pore size

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
40161507
NACRES:
NB.24
eCl@ss:
32031602
Pore size:
0.22 μm pore size, 75 % porosity
Material:
mixed cellulose esters (MCE) membrane, plain filter, white filter
Feature:
hydrophilic
Diam.:
142 mm
Sterility:
non-sterile
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

material

mixed cellulose esters (MCE) membrane, plain filter, white filter

description

142 mm diameter, mixed cellulose esters (MCE) membrane, hydrophilic, white, 50 discs

sterility

non-sterile

feature

hydrophilic

manufacturer/tradename

MF-Millipore, Millipore

parameter

18 mL/min-cm2 water flow rate, 2 L/min-cm2 air flow rate, 75 °C max. temp.

diam.

142 mm

filter diam.

142 mm

thickness

150 μm

gravimetric extractables

2%

color

white

refractive index

n/D 1.51

matrix

MF-Millipore

pore size

0.22 μm pore size, 75 % porosity

capacity

150 μg/cm2 binding capacity (protein)

bubble point

≥3.52 bar, air with water at 23 °C

shipped in

ambient

General description

Biologically inert mixtures of cellulose acetate and cellulose nitrate have made MF-Millipore membrane filters one of the most widely used membranes in analytical and research applications.

MF-Millipore filters without Triton surfactant contain minimum amounts of wetting agent and have a lower water extractable content than standard MF-Millipore filters.

Features & Benefits:
- Versatile filter for biological and environmental monitoring applications
- Available in a range of pore sizes, colored black or white, with or without a gridded surface
- Compatible with ethylene oxide, gamma irradiation, and autoclave sterilization methods
Filter Code: GSWP

Application

  • Sterilizing filtration
  • Bioassays
  • Isolation of virus-like particles in wastewater
  • Microplastics analysis grade water
  • Nucleic acid binding, including eDNA

Other Notes

Directions for Use

  • Organism Retention: Microorganism
  • Mode of Action: Filtration (size exclusion)
  • Application: General laboratory filtration
  • Intended Use: Retention or removal of biological contaminants
  • Instructions for Use: Sterilizing filtration of a liquid through a membrane with a 0.2 μm (or smaller) pore size effectively removes biological contaminants, including bacteria, mold and yeast. For the selective retention of larger biological contaminents, liquid filtration through membranes with 0.45 μm (or larger) pore sizes may be used to trap and support microorganism growth for subsequent culture and analysis
  • Storage Statement: Store in dry location away from heat source
  • Disposal Statement: Dispose of in accordance with applicable federal, state and local regulations.

Legal Information

MF-Millipore is a trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?


pictograms

Flame

signalword

Danger

Storage Class

4.1B - Flammable solid hazardous materials

Hazard Classifications

ED ENV 1 - Flam. Sol. 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library