Skip to Content
MilliporeSigma

GR11

Anti-IGF-IR (Ab-1) Mouse mAb (αIR3)

liquid, clone αIR3, Calbiochem®

Synonym(s):

Anti-IGF-R, Anti-Insulin-Like Growth Factor Receptor, Anti-Insulin-Like Growth Factor Receptor, Anti-IGF-R

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.43
UNSPSC Code:
12352203
Clone:
αIR3, monoclonal
Species reactivity:
human
Application:
Citations:
10
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

antibody form

purified antibody

antibody product type

primary antibodies

clone

αIR3, monoclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

dilution

(Immunoblotting (not recommended)
Immunofluorescence (1-5 µg/mL)
Immunoprecipitation (1-2 µg/sample)
Neutralization Studies (1 µg/mL)
Paraffin Sections (not recommended))

isotype

IgG1

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... IGF1R(3480)

General description

Purified mouse monoclonal antibody generated by immunizing SJL mice with the specified immunogen and fusing splenocytes with FO mouse myeloma cells (see application references). Recognizes the ~130 kDa α and the ~90 kDa β subunits of IGF-I receptor.
Recognizes the ~130 kDa α and the ~90 kDa β subunits of IGF-I receptor.
This Anti-IGF-IR (Ab-1) Mouse mAb (αIR3) is validated for use in Immunoblotting, IF, IP, Neutralization Studies, Paraffin Sections for the detection of IGF-IR (Ab-1).

Immunogen

Human
partially purified, human placenta IGF-I receptor

Application

Immunoblotting (not recommended)

Immunofluorescence (1-5 µg/ml)

Immunoprecipitation (1-2 µg/sample)

Neutralization Studies (1 µg/ml; use Cat. No. GR11L)

Paraffin Sections (not recommended)

Packaging

Please refer to vial label for lot-specific concentration.

Physical form

In 0.05 M sodium phosphate buffer, 0.2% gelatin.

Analysis Note

Negative Control
HS27 cells
Positive Control
HepG2 cells

Other Notes

Anti-IGF-I Receptor (Ab-1) Mouse mAb (αIR3) immunoprecipitates the α and β subunits of the IGF-I receptor. Blocks IGF-I binding to its receptor and may bind weakly to the insulin receptor. It also inhibits the growth of MCF-7 cells in culture suggesting the IGF-I receptor may be involved in autocrine regulation of cell growth (see application references). Antibody should be titrated for optimal results in indvidual systems.
Roth, R. 1988. Science239, 1269.
Rohlik, Q.T., et al. 1987. Biochem. Biophy Res. Comm.149, 276.
Rosen, O.M., 1987. Science257, 1452.
Rechler, M.M. and Nissley, S.P., 1986. Hormone Res.24, 152.
Ullrich, A., et al. 1986. EMBO J.5, 2503.
Zapf, S. and Froesch, E.R., 1986. Hormone Res.24, 121.
Humbel, R.E., 1984. I. Chemistry; in Li Hormonal proteins and peptides. Vol 12, Chap. 4 (Academic Press, New York).
Kull, F.C., et al. 1983. J. Biol. Chem.258, 6561.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Still not finding the right product?

or

Try our Product Selector Tool to narrow your options


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library