Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GF333

Sigma-Aldrich

IL-2,
Human Recombinant Animal Free

Synonym(s):

Interleukin-2, IL2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.77

impurities

<0.1 μg/mg protein endotoxin (<1 EU/μg)

Quality Level

UniProt accession no.

shipped in

wet ice

General description

IL-2 is a powerful immunoregulatory lymphokine produced by T-cells in response to antigenic or mitogenic stimulation. IL-2/IL-2R signaling is required for T-cell proliferation and other fundamental functions which are essential for the immune response. IL-2 stimulates growth and differentiation of B-cells, NK cells, lymphokine activated killer cells, monocytes, macrophages and oligodendrocytes.
Recombinant human IL-2 is a 15.5kDa protein, containing 134 amino acid residues including one intrachain disulfide bond.
Product Source: Protein is expressed in E.coli.
The protein is manufactured without the use of any animal products/reagents and is considered Animal-Free and can be used in protocols where possible transmisson of animal viruses and contamination must be avoided.

Application

Research Category
Stem Cell Research
Research Sub Category
Growth Factors & Receptors

Quality

Determined its its ability to stimulate the proliferation of mouse CTLL-2 cells. The expected ED50 is ≤ 0.1 ng/ml, corresponding to a specific activity of ≥ 1 x 10^7 units/mg.

Physical form

Product is presented without additives and is filtered through a 0.2 micron filter before lyophilization.

Storage and Stability

Store at -20°C for up to 4 months from date of receipt Centrifuge the vial prior to opening.
Reconstitute in 100mM Acetic Acid to a concentration of 0.1-1.0 mg/ml. Do not vortex. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein and store in working aliquots at -20°C.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn about the impact of filtration on hematopoietic cell culture media containing vital T cell factors and other media components.

Learn about the impact of filtration on hematopoietic cell culture media containing vital T cell factors and other media components.

Learn about the impact of filtration on hematopoietic cell culture media containing vital T cell factors and other media components.

Learn about the impact of filtration on hematopoietic cell culture media containing vital T cell factors and other media components.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service