Skip to Content
MilliporeSigma
All Photos(1)

Documents

GF301

Sigma-Aldrich

BDNF Protein, Human Recombinant Animal Free

The BDNF protein is a member of the NGF family of neurotrophic factors (also named neurotrophins) that are required for the differentiation & survival of specific neuronal subpopulations in both the central as well as the peripheral nervous system.

Synonym(s):

Brain derived neurotrophic factor, Abrineurin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.77

Quality Level

Assay

>98% (SDS-PAGE and HPLC)

impurities

<0.1 μg/mg protein endotoxin (<1EU/μg)

UniProt accession no.

shipped in

dry ice

General description

Brain derived neurotrophic factor (BDNF) is a member of the NGF family of neurotrophic factors (also named neurotrophins) that are required for the differentiation and survival of specific neuronal subpopulations in both the central as well as the peripheral nervous system. The neurotrophin family is comprised of at least four proteins including NGF, BDNF, NT3, and NT4/5. BDNF mediates its neurotrophic properties by signaling through a high affinity cell surface receptor called gp145/trkB. BDNF is expressed as the C-terminal portion of a 247 amino acid polypeptide precursor, which also contains a signal sequence of 18 amino acid residue and a propeptide of 110 amino acid residues. Recombinant human BDNF is a 27.0 kDa homodimer of two 119 amino acid subunits linked by strong non-covalent interactions.
Product Source: E. coli

Specificity

Cross Reactivty
Chicken, Frog, Mouse, Rat, Hamster, Guinea Pig, and Monkey

Application

Research Category
Neuroscience
Research Sub Category
Growth Factors & Receptors

Quality

Bioactivity assay: Determined by its ability to stimulate the proliferation of rat C6 cells. The expected ED50 for this effect is 0.5-1.0 μg/ml.

Physical form

Product is filtered through a 0.2 micron filter before lyophilization.
Recombinant animal free human BDNF is manufactured using all non-animal reagents

Storage and Stability

Store at -20°C for up to 6 months from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service