Skip to Content
MilliporeSigma

GF141

Stem Cell Factor Protein, Recombinant mouse

The Stem Cell Factor (SCF) Protein, Recombinant mouse, is a hematopoietic growth factor that exerts its activity at the early stages of hematopoiesis.

Synonym(s):

SCF, Mast Cell Growth Factor, MGF, c-kit Ligand

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77
eCl@ss:
32160405
Assay:
>98% (SDS-PAGE)
Biological source:
mouse
Recombinant:
expressed in E. coli
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

recombinant

expressed in E. coli

assay

>98% (SDS-PAGE)

specific activity

≥1x10e5 U/mg

manufacturer/tradename

Chemicon®

impurities

<0.1 ng/μg Endotoxin (of SCF)

input

sample type hematopoietic stem cell(s)

UniProt accession no.

shipped in

dry ice

storage temp.

-10 to -25°C

Gene Information

mouse ... KITLG(17311)

General description

Product Source: E. coli
Stem Cell Factor (SCF) is a hematopoietic growth factor that exerts its activity at the early stages of hematopoiesis. SCF stimulates the proliferation of myeloid, erythroid, and lymphoid progenitors in bone marrow cultures and has been shown to act synergistically with colony stimulating factors. Murine SCF is an 18.3 kDa protein containing 164 amino acid residues.

Application

Concentration Range for most in vitro applications is 1.0 - 100 ng/mL. Optimal working dilutions must be determined by end user.
The Stem Cell Factor (SCF) Protein, Recombinant mouse, is a hematopoietic growth factor that exerts its activity at the early stages of hematopoiesis.

Physical form

Lyophilized protein from 10 mM Sodium Phosphate, pH 6.5, containing no additives.

Preparation Note

The lyophilized powder, though stable at room temperature, is best stored at -20°C for up to 12 months from date of receipt. After a quick spin, reconstitute with sterile water to a concentration of 0.1-1.0 mg/mL. This solution can be diluted into other buffered solutions and stored at 4°C for one week or at -20°C for future use. Reconstituted mouse SCF should be stored in working aliquots at -20°C for up to six months. Avoid repeated freeze/thaw cycles.

Analysis Note

Specific Activity: Murine SCF is biologically active when compared to standards.The ED(50), as determined by the dose-dependent stimulation of the proliferation of the human TF-1 cell line is ≤ 10 ng/mL

Other Notes

Replaces: GF049

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Storage Class

11 - Combustible Solids

wgk

WGK 2



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library