Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
12352203
NACRES:
NA.44
eCl@ss:
32160702
Conjugate:
FITC conjugate
Clone:
AT13/5, monoclonal
Application:
FACS
Citations:
-
biological source
mouse
Quality Segment
conjugate
FITC conjugate
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
AT13/5, monoclonal
species reactivity
human
manufacturer/tradename
Milli-Mark®
technique(s)
flow cytometry: suitable
isotype
IgG1κ
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... CD38(952)
General description
45 kDa (Deaglio, 2001)
CD38 is a 45 kDa type II integral membrane glycoprotein consisting of an N-terminal 20 amino acid cytoplasmic domain, a 23 amino acid transmembrane domain, and a 257 amino acid extracellular domain. CD38 is associated with different molecules in different cell types and these unique combinations cause unique cell-type specific signaling. One of the known ligands that activates CD38-mediated signaling is CD31 (Deaglio, 2001). CD38 is expressed by a number of different cell types and the pattern of expression often changes with differentiation and maturation. Thus, CD38 expression is found during early stages of B and T-cell differentiation, is lacking during the later intermediate stages, and is regained near the end of maturation. CD38 is expressed at variable levels on the majority of hematopoietic cells, prevalently during early differentiation and activation, and it is expressed strongly on plasma cells (Jackson, 1990; Terstappen, 1990; Morra, 1997). CD38-postive cell types outside the hematopoietic system include subsets from the gut, brain, prostate, pancreas, bone, eye, and muscle (Deaglio, 2001). CD38 is expressed on approximately 60% of CD34-positive peripheral blood mononuclear cells. The least mature CD34-postive cells are characterized by a lack of CD38 (Herbein, 1994).
Anti-CD38, AT13/5 was included in the Sixth International Workshop and Conference on Human Leucocyte Differentiation Antigens, and studies by a number of laboratories confirmed its reactivity with CD38 (Morra, 1997).
Anti-CD38, AT13/5 was included in the Sixth International Workshop and Conference on Human Leucocyte Differentiation Antigens, and studies by a number of laboratories confirmed its reactivity with CD38 (Morra, 1997).
Immunogen
Namalwa cells (B lymphocyte cell line from Burkitt′s lymphoma)
Application
Milli-Mark Anti-CD38-FITC Antibody, clone AT13/5 is an antibody against CD38 for use in FC.
Research Category
Inflammation & Immunology
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology
Immunoglobulins & Immunology
Biochem/physiol Actions
Antibody recognizes Human CD38 antigen.
Physical form
Protein A purified
Purified Mouse monoclonal IgG1 conjugated to FITC in 0.05M Tris-HCl buffer with 1% BSA, Sodium Azide at 0.097% at pH 7.2.
Preparation Note
Maintain refrigerated at 2-8°C protected from light in undiluted aliquots for up to 6 months from date of receipt.
Analysis Note
Control
Human Lymphocytes
Human Lymphocytes
Evaluated by flow cytometry using human lymphocytes
Legal Information
MILLI-MARK is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.