Skip to Content
MilliporeSigma
All Photos(1)

Documents

FCMAB185AP

Sigma-Aldrich

Milli-Mark® Anti-CD19 -APC Antibody, clone HD37

clone HD37, Milli-Mark®, from mouse

Synonym(s):

Differentiation antigen CD19

B-lymphocyte surface antigen B4

T-cell surface antigen Leu-12, B-lymphocyte antigen CD19

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

Allophycocyanin conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

HD37, monoclonal

species reactivity

human

manufacturer/tradename

Milli-Mark®

technique(s)

flow cytometry: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CD19(930)

General description

CD19 is a 120 kDa transmembrane glycoprotein with a reduced MW of 95 kDa (Pezzutto, 1987; Sato, 1997). CD19 is a member of the immunoglobulin superfamily with two extracellular C2-type domains (Pezzutto, 1987), and it is a critical signal transduction molecule that regulates B lymphocyte development, activation, and differentiation (Sato, 1997). CD19 expression is restricted to B cells, being absent from T cells, monocytes, and granulocytes (Pezzutto, 1987). The CD19 antigen appears early during B cell maturation, probably at late pro-B cell stage around the time of Ig heavy chain rearrangement. It then persists during all stages of B cell maturation and is lost upon terminal differentiation to plasma cells (Pezzutto, 1987).

Specificity

Antibody recognizes Human CD19 antigen.

Immunogen

Hairy cell leukemia cells

Application

Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology
This Milli-Mark Anti-CD19 -APC Antibody, clone HD37 is validated for use in FC for the detection of CD19 -APC.

Quality

Evaluated by flow cytometry using Human Lymphocytes

Target description

120 kDa (Pezzutto, 1987; Sato, 1997)

Physical form

Protein A purified
Purified Mouse monoclonal IgG1 conjugated to APC in 0.05M Tris-HCl buffer with 1% BSA, Sodium Azide at 0.097% at pH 7.2

Storage and Stability

Maintained refrigerated at 2-8°C from date of receipt. Protected from light in undiluted aliquots for 6 months from date of receipt.

Analysis Note

Control
Human Lymphocytes

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

MILLI-MARK is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service