Skip to Content
MilliporeSigma

FC014

Bovine Fibronectin

from bovine plasma, liquid, suitable for cell culture

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.75
Biological source:
bovine
Form:
liquid
Technique(s):
cell culture | mammalian: suitable
Concentration:
1.0 mg/mL
Assay:
~95% (affinity purified, SDS-PAGE)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Fibronectin, Bovine,

biological source

bovine

Quality Level

assay

~95% (affinity purified, SDS-PAGE)

form

liquid

manufacturer/tradename

Chemicon®

concentration

1.0 mg/mL

technique(s)

cell culture | mammalian: suitable

input

sample type induced pluripotent stem cell(s)
sample type mesenchymal stem cell(s)
sample type: mouse embryonic stem cell(s)
sample type epithelial cells
sample type pancreatic stem cell(s)
sample type neural stem cell(s)
sample type hematopoietic stem cell(s)

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... FN1(2335)

Looking for similar products? Visit Product Comparison Guide

General description

Bovine fibronectin was purified from bovine plasma over a gelatin agarose column. A single band at 220kDa (reduced) is identified, corresponding to intact fibronectin under reduced conditions.
Product Source: Bovine plasma

Application

ELISA, Ouchterlony and immunoblotting.

Physical form

Liquid in a solution of 2M Urea in 0.05 M Tris pH 7.0.

Preparation Note

Aliquot and store up to 12 months at -20°C. For short term (1-2 weeks) use, storage at 2-8°C is acceptable. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Ruoslahti et al.
The Biochemical journal, 193(1), 295-299 (1981-01-01)
Human fibronectin was isolated from second-trimester amniotic fluid, from amniotic fluid obtained at term and from adult plasma. The amniotic-fluid fibronectins had a slightly higher apparent molecular weight on sodium dodecyl sulphate/polyacrylamide-gel electrophoresis than the plasma fibronectin. Early- and late-amniotic-fluid

Global Trade Item Number

SKUGTIN
FC01404053252269929

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service