Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
12352104
Quality Segment
form
liquid
composition
Ethanol, 70 % (w/v) , Water, 28-32 % (v/v)
color
colorless
transition temp
flash point 10.6 °C
density
0.8750-0.8850 g/mL at 20 °C
format
mixture
shipped in
ambient
storage temp.
room temp
General description
EMPARTA solvents are designed for routine chemical synthesis, drying, purification, and critical labware cleaning in quality control environments. Engineered for precision, they deliver consistently reproducible results essential for rigorous validation and documentation.
Exceptional Quality Standards: These solvents meet or exceed ACS specifications, providing assurance of high quality that is crucial for accurate testing and analysis in quality control.
Consistent Reproducibility: Designed for precision, the solvents ensure that results can be reliably reproduced, which is vital for maintaining compliance and integrity in quality control processes.
Versatile Applications: Suitable for a variety of QC tasks, including analytical testing, method validation, and testing USP/NF monographs, these solvents support a comprehensive approach to quality assurance in laboratory settings.
Exceptional Quality Standards: These solvents meet or exceed ACS specifications, providing assurance of high quality that is crucial for accurate testing and analysis in quality control.
Consistent Reproducibility: Designed for precision, the solvents ensure that results can be reliably reproduced, which is vital for maintaining compliance and integrity in quality control processes.
Versatile Applications: Suitable for a variety of QC tasks, including analytical testing, method validation, and testing USP/NF monographs, these solvents support a comprehensive approach to quality assurance in laboratory settings.
Application
Ethanol Solution 70% W/V (in H2O) can be used as:,·
- An extraction solvent for the extraction of phenolic compounds.
- A dehydrating reagent in tissue processing protocol.
Packaging
M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!
Analysis Note
Signal Word
Danger
Hazard Codes
Precautionary Statements
Hazard Classifications
Carc. 2 Inhalation - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 2
Target Organs
Eyes,Central nervous system
Storage Class
3 - Flammable liquids
WGK
WGK 1
Flash Point (°F)
51.1 °F
Flash Point (°C)
10.6 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


