Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

EFHAW250

Millipore

EZ-Fit® filtration unit with absorbent pad

pore size 0.45 μm, mixed cellulose esters (MCE) membrane, white filter, volume 250 mL, pkg of Single packaging (EFHAW250I), pkg of Bulk packaging (EFHAW250B), suitable for (suitable for solid and liquid cultivation)

Synonym(s):

EZ-Fit® Filtration Unit, Membrane filtration device made of gridded, mixed cellulose esters membrane for bioburden testing

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
NB.24

material

gridded filter
mixed cellulose esters (MCE) membrane
white filter

Quality Level

Agency

ISO 7004

description

General microbiological analysis

sterility

electron beam irradiated

feature

ready-to-use

manufacturer/tradename

EZ-Fit®

packaging

pkg of Bulk packaging (EFHAW250B)
pkg of Single packaging (EFHAW250I)

filter diam.

47 mm

funnel capacity

100 mL

volume

250 mL

pore size

0.45 μm pore size

input

sample type pharmaceutical(s)
sample type beverage(s)
sample type food(s)

suitability

suitable for (suitable for solid and liquid cultivation)

application(s)

bioburden testing
cosmetics
food and beverages
pharmaceutical
water monitoring

compatibility

for use with EZ-Fit® System

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

The EZ-FIT® filtration unit is a single-use filtration device made of white, gridded, and mixed cellulose esters (MCE) membrane with a blue base integrating a cellulose support pad. It provides a safe and fast way of performing membrane filtration by avoiding the exposure to the risks of microbiological analysis.

The filtration units are e-beam irrated, a vented lid covers the sample throughout filtration, and a 360° rim prevents forceps from accidentally touching the membrane filtration area during membrane transfer, to protect samples from contamination while ensuring reliable microbial enumeration results.

The EZ-FIT® Azure feature makes the EZ-FIT® filtration unit even more ergonomic and convenient with transparent funnel and all-around filling level indicators for an easy filling from any angle. The innovative funnel assembly design prevents any leaks or sample by-pass of the membrane.

The blue units provide the extra flexibility of either transferring the membrane to an agar plate or converting the device into a Petri dish by adding the liquid media, after filtration.

The filtration units conform to international standards and minimize sample residue. The units can be stacked to save the work area. This disposable device is designed to fit onto the EZ-FIT® Manifold, which features specific EZ-FIT® Filtration heads.

Application

The EZ-FIT® filtration units are used in microbiological analysis and bioburden testing of liquid samples such as in process and final product samples in beverages, pharmaceuticals and water. These products are also used in industrial QC and water monitoring.

Packaging

EFHAW250I: Individual packaging (48 units/pack)
EFHAW250B: Bulk packaging with protective bag (48 units/pack)

Legal Information

EZ-FIT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Comparison of convenience, operating time, accuracy, costs and safety aspects of single-use funnels vs. reusable funnels for the membrane filtration method in QC microbiology.

Comparison of convenience, operating time, accuracy, costs and safety aspects of single-use funnels vs. reusable funnels for the membrane filtration method in QC microbiology.

Comparison of convenience, operating time, accuracy, costs and safety aspects of single-use funnels vs. reusable funnels for the membrane filtration method in QC microbiology.

Comparison of convenience, operating time, accuracy, costs and safety aspects of single-use funnels vs. reusable funnels for the membrane filtration method in QC microbiology.

See All

Related Content

Clean drinking water is essential for food and beverage production. This article gives an overview of chemical and microbiological testing of the drinking water for quality and safety.

Clean drinking water is essential for food and beverage production. This article gives an overview of chemical and microbiological testing of the drinking water for quality and safety.

Clean drinking water is essential for food and beverage production. This article gives an overview of chemical and microbiological testing of the drinking water for quality and safety.

Clean drinking water is essential for food and beverage production. This article gives an overview of chemical and microbiological testing of the drinking water for quality and safety.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service