Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ECM671

Sigma-Aldrich

QCM Gelatin Invadopodia Assay (Red)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.76

technique(s)

cell migration: suitable

Quality Level

detection method

fluorometric

Application

The EMD Millipore QCM Gelatin Invadopodia Assay (Red) provides the reagents necessary for affixing a thin, uniform layer of Cy3-labeled gelatin to a glass culture substrate, allowing for rapid detection of matrix degradation (Artym et al., 2009, Xu et al., 2009). A poly-L-lysine coating is first adsorbed to the glass substratum. The substrate is then treated with a dilute glutaraldehyde solution to bi-functionally “activate” the surface for further protein binding. Subsequent incubation of the surface with fluorescent gelatin allows covalent coupling between the poly-L-lysine and gelatin via reactive aldehyde (-CHO) groups. The fluorescently-coated glass is now prepared for cell culture by disinfection with 70% ethanol, followed by quenching of free aldehydes with amino acid-containing growth medium. Upon completion of fluorescent substrate preparation, cell types of interest may be seeded onto the gelatin surface for a desired amount of time. Depending on cell type, degradation may occur within a few to several hours. Treatment compounds of interest may also be introduced within the culture period.

With EMD Millipore′s QCM Gelatin Invadopodia Assay (Red), degraded areas of gelatin, now devoid of fluorescence, may be microscopically visualized and quantified using image analysis software algorithms. The assay also provides fluorescent FITC-phalloidin and DAPI, for visualization of cytoskeletal F-actin and nuclei, respectively, to allow co-localization of degradation with cellular features. Potential activators or inhibitors of invadopodia formation may be investigated for their influence on the degree and frequency of matrix degradation, and the assay may be further combined with immunocytochemical staining for other molecules of interest in pathway studies.

Packaging

Enough Reagents for 4 x 8-well Chamber Slides (32 assays)

Components

Cy3-Gelatin, 4X: One vial containing 0.5 mL Unlabeled Gelatin, 4X: One vial containing 2 mL DAPI: One vial containing 100 µL at 100 µg/mL in water Poly-L-Lysine, 2X: One vial containing 5 mL Glutaraldehyde, 16X: One vial containing 1 mL FITC-Phalloidin: One vial containing 20 µg

Storage and Stability

Store Cy3-Gelatin, Unlabeled Gelatin, Poly-L-Lysine and DAPI at 2-8°C. Store Glutaraldehyde and FITC-Phalloidin at -20°C. Use all reagents within 4 months from date of receipt.

Legal Information

Cy3 is a trademark of Cytiva

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service