Skip to Content
MilliporeSigma

ECM104

Millicoat® Human Collagen Type I Coated Strips (96-Wells)

96-well plate coated with human Collagen Type I used for cell adhesion studies.

Synonym(s):

Formerly under the CytoMatrix ™ brand name.

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352202
eCl@ss:
32161000
NACRES:
NA.75
Biological source:
human
Form:
lyophilized
Technique(s):
activity assay: suitable, cell based assay: suitable
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human

Quality Segment

form

lyophilized

species reactivity

human

manufacturer/tradename

Chemicon®, Millicoat®

technique(s)

activity assay: suitable, cell based assay: suitable

input

sample type mesenchymal stem cell(s)
sample type neural stem cell(s)
sample type epithelial cells
sample type hematopoietic stem cell(s)
sample type induced pluripotent stem cell(s)
sample type: mouse embryonic stem cell(s)
sample type: human embryonic stem cell(s)
sample type pancreatic stem cell(s)

NCBI accession no.

UniProt accession no.

detection method

colorimetric

shipped in

wet ice

storage temp.

2-8°C

Gene Information

human ... COL1A1(1277)

General description

Cell adhesion plays a major role in cellular communication and regulation, and is of fundamental importance in the development and maintenance of tissues. Scientists are continually examining the adhesion and migration of many diverse cell types on various extracellular matrix (ECM) component proteins. Millicoat Cell Adhesion Strips are provided as 12 x 8-well removable strips in a plate frame for convenience and flexibility in designing assays. The wells in rows A - G have been coated with Human Collagen Type I. Row H of each strip is coated with BSA which serves as a negative assay control. Cells are seeded onto the coated substrate. Subsequently, adherent cells are fixed and stained. Relative attachment is determined using absorbance readings.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLICOAT is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library