Skip to Content
MilliporeSigma
All Photos(3)

Documents

E711

Millipore

NovaSeptum® GO High Purity Bag Sampling System

sterile; β-irradiated

Synonym(s):

NovaSeptum® GO High Purity Bag, Sterile_Sampling_Container

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104105

material

316 stainless steel cannula (ASTM®)
PureFlex bag
TPE tubing (Thermoplastic elastomer)
polyester body
silicone septum (platinum-cured)

Quality Level

Agency

according to ISO 11137 (sterilization)
according to ISO 146441
meets USP 88 biological reactivity (all component materials)

sterility

sterile; β-irradiated

product line

NovaSeptum® GO

feature

autoclavable: no

parameter

-20-50 °C temp. range (-4-122 °F) (for 100 mL)
-80-50 °C temp. range (-112-122 °F) (for 50 mL to 250 mL (when used with a NovaSeptum® Case))
0.30 bar max. pressure (4.35 psi) (for 1000 mL)
0.50 bar max. pressure (7.25 psi) (up to 250 mL
0.30 bar (4.35 psi) for 1000 mL)

impurities

<2.15 EU/device bacterial endotoxins (LAL test)

fitting

female Luer-Lok® outlet (3-piece )
male Luer-Lok® outlet (3-piece containing an injection site)

application(s)

cell therapy
gene therapy
life science and biopharma
mAb
ophthalmics
parenterals
pathogen testing
pathogen testing
pharma/biopharma processes
pyrogen testing
sterile sampling
sterility testing
viral therapy

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

A closed, sterile sampling method can significantly reduce risk of cross contamination. Ideal for sampling from aseptic and sterile processes, the NovaSeptum® GO sterile sampling system provides consistently representative samples, safely and securely. NovaSeptum® GO high purity bags for single sampling systems are available from 50 to 1000 mL.
Device configuration: single Unit Sampling

Application

Regular Sampling Sample (with or without protective case).
Sterility testing, pyrogenic testing, endotoxin testing, chemical analysis, pH analysis, and fermentation applications.

Packaging

E711-10050 (50 ml bags, 50 units per box)
E711-10100 (100 ml bags, 50 units per box)
E711-10250 (250 ml bags, 50 units per box)
E711-11000 (1000 ml bags, 50 units per box)

Components

  • Septum: Platinum-cured silicone
  • Body: Polyester
  • Cannula: ASTM® 316 L Stainless steel
  • Fluid contact layer (bag): Polyethylene film (PureFlex film)
  • Tubing: Thermoplastic elastomer (TPE)
  • Outlet Tubing: 3-piece Luer-Lok®, containing a male, female and an injection site

Preparation Note

Units are integrity tested at regular intervals during manufacturing.
Aqueous extraction contains < 2.15 EU per device as determined using the Limulus Amebocyte Lysate (LAL) test.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials
Luer-Lok is a registered trademark of Becton-Dickinson & Co.
NOVASEPTUM is a registered trademark of Merck KGaA, Darmstadt, Germany
PUREFLEX is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

A cost-model compares modalities like viral vectors and mRNA for production scale decision-making.

A cost-model compares modalities like viral vectors and mRNA for production scale decision-making.

A cost-model compares modalities like viral vectors and mRNA for production scale decision-making.

A cost-model compares modalities like viral vectors and mRNA for production scale decision-making.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service