Skip to Content
MilliporeSigma
All Photos(1)

Documents

DR1088

Sigma-Aldrich

Anti-TWIST1 Rabbit pAb

liquid, Calbiochem®

Synonym(s):

Anti-TWIST Homolog 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

does not contain

preservative

species reactivity

human, mouse, rat

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TWIST1(7291)
mouse ... Twist1(22160)

General description

Purified rabbit polyclonal antibody. Recognizes the ~25 kDa TWIST1 protein.
Recognizes the ~25 kDa TWIST1 protein in HeLa cells.
This Anti-TWIST1 Rabbit pAb is validated for use in Immunoblotting for the detection of TWIST1.

Immunogen

Full-length, human TWIST1 (aa 1-202)
Human

Application

Immunoblotting (1-5 µg/ml)

Warning

Toxicity: Regulatory Review (Z)

Physical form

In PBS, pH 7.2.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Negative Control
293T cells
Positive Control
HeLa cells

Other Notes

Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service