Skip to Content
MilliporeSigma

CBL496F

Anti-CD34 Class II Antibody, clone QBEND/10, FITC conjugated

clone QBEND/10, Chemicon®, from mouse

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
FITC conjugate
Clone:
QBEND/10, monoclonal
Application:
FACS
Citations:
1
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

FITC conjugate

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

QBEND/10, monoclonal

species reactivity

monkey, human

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CD34(947)

Immunogen

Human endothelial vesicles

Application

Identification of haemopoietic progenitor cells

Studies into the control mechanisms of early haemopoiesis

Purification of haemopoietic cells from, for example, FACS

The antibody reacts with both frozen and fixed tissue sections from vascular endothelium and stromal fibroblasts

Optimal working dilutions must be determined by the end user.
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology
This Anti-CD34 Antibody (Class II), clone QBEND/10, FITC conjugated is validated for use in FC for the detection of CD34 Class II.

Biochem/physiol Actions

The antibody recognizes a heavily glycosylated transmembrane protein: gp 105-120 kDa. The antigen is expressed on immature human haemopoietic precursor cells, capillary endothelial cells and human myeloid cells in the following categories: subsets of CFU-GEMM and BFU-E, all CFU-GM, granulocyte precursors, monocytes, myeloid leukaemias. This antibody recognizes the class II epitope showing resistance to neuraminidase enzyme cleavage.

FUSION PARTNER: NSO myeloma cell line

Physical form

The conjugate is supplied as a 100 test vial in phosphate buffered saline containing 10mM sodium azide and 1mg/ml bovine serum albumin. For flow cytometry we recommend using 10μL of conjugate per test.

Preparation Note

Store at +4°C protected from light. DO NOT FREEZE. For long term use and storage aliquot conjugate into small volumes and store at +4°C.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library