Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CBL2117

Sigma-Aldrich

Anti-Interleukin-6 Antibody, clone B-E8

clone B-E8, Chemicon®, from mouse

Synonym(s):

IL-6, B-cell stimulatory factor 2, Interferon beta-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

100
300

antibody form

purified antibody

antibody product type

primary antibodies

clone

B-E8, monoclonal

species reactivity

human

should not react with

mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
flow cytometry: suitable
immunohistochemistry: suitable
neutralization: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

Gene Information

human ... IL6(3569)

Specificity

Recognizes natural and recombinant forms of human Interleukin-6 with an affinity of 0.5 nM. Inhibits IL-6 binding to its receptors and blocks Interleukin-6 induced proliferation on B9 cells.

Immunogen

Recombinant human IL-6.

Application

Blocks Interleukin-6 induced proliferation on B9 cells: 20pg of antibody neutralizes the activity of 1U of IL-6. It is recommended that the sodium azide is removed from the antibody prior to use in this application.

Immunohistochemistry on frozen sections

Optimal working dilutions must be determined by the end user.
Detect Interleukin-6 using this Anti-Interleukin-6 Antibody, clone B-E8 validated for use in ELISA, FC, IH, NEUT.
Research Category
Inflammation & Immunology
Research Sub Category
Cytokines & Cytokine Receptors

Linkage

Replaces: 04-1095

Physical form

Format: Purified
The monoclonal is purified from tissue culture supernatant by ion exchange chromatography and is presented as a liquid in phosphate buffered saline, pH 7.2.

Storage and Stability

Store at 2-8°C for up to 12 months from date of receipt. For long term storage aliquot antibody into small volumes and store at -20°C. Avoid repeated freeze-thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Microglia derived IL-6 suppresses neurosphere generation from adult human retinal cell suspensions.
Balini Balasubramaniam,Debra A Carter,Eric J Mayer,Andrew D Dick
Experimental Eye Research null
Elevated levels of IL-18 in plasma and skeletal muscle in chronic obstructive pulmonary disease.
A M W Petersen,M Penkowa,M Iversen,L Frydelund-Larsen,J L Andersen,J Mortensen,P Lange,B K Pedersen
Lung null
Immunohistochemical detection of interleukin-6 in human skeletal muscle fibers following exercise.
Penkowa, M; Keller, C; Keller, P; Jauffred, S; Pedersen, BK
Faseb Journal null
Berberine inhibits inflammatory response and ameliorates insulin resistance in hepatocytes.
Tianjiong Lou,Zhongai Zhang,Zhilei Xi,Kang Liu,Lin Li,Baolin Liu,Fang Huang
Inflammation null
Zifeng Liu et al.
Journal of diabetes, 8(3), 324-335 (2015-04-09)
Resveratrol is a phytoalexin with beneficial effects on human health. The aim of the present study was to investigate the effects of resveratrol on endothelial dysfunction involved in insulin signaling and inflammation. Endothelial cells were stimulated with palmitate (PA) to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service