Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CBL197

Sigma-Aldrich

Anti-Cytokeratin 14 Antibody, clone LL002

clone LL002, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

LL002, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG3

NCBI accession no.

UniProt accession no.

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... KRT14(3861)

Application

Anti-Cytokeratin 14 Antibody, clone LL002 is an antibody against Cytokeratin 14 for use in WB, IH(P).
Suitable for staining formalin fixed wax sections or frozen tissues. In order to obtain 100% consistency in staining it is considered essential to either microwave treat fixed tissue sections or alternatively use the more recently recommended high temperature antigen unmasking technique. Prior exposure of fixed tissues to enzyme (trypsin) digestion is deleterious to epitope expression. The fixative recommended for use with CBL 197 is neutral buffered formalin.

Western blotting

Optimal working dilutions must be determined by the end user.

Quality

Tested

Physical form

Format: Purified
The monoclonal is presented at a concentration of 100 μg/mL in phosphate buffered saline containing 10mM sodium azide and 1% bovine serum albumin. We recommend that each laboratory determine an optimum working titre for use in its particular application.

Other Notes

Please refer to lot specific datasheet.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Caitlynn M L Barrows et al.
Methods in molecular biology (Clifton, N.J.), 2749, 25-38 (2023-12-22)
Resident stem/progenitor cells within the secretory salivary glands offer a potential therapeutic resource for use in the regeneration of salivary glands needed to restore saliva production in patients with chronic xerostomia, or dry mouth. Methods were developed previously to isolate
Sara Nagosa et al.
Stem cell reports, 9(6), 1991-2004 (2017-12-05)
miR-184 is a highly evolutionary conserved microRNA (miRNA) from fly to human. The importance of miR-184 was underscored by the discovery that point mutations in miR-184 gene led to corneal/lens blinding disease. However, miR-184-related function in vivo remained unclear. Here, we
Gernot Neumayer et al.
Nature communications, 15(1), 5834-5834 (2024-07-12)
We present Dystrophic Epidermolysis Bullosa Cell Therapy (DEBCT), a scalable platform producing autologous organotypic iPS cell-derived induced skin composite (iSC) grafts for definitive treatment. Clinical-grade manufacturing integrates CRISPR-mediated genetic correction with reprogramming into one step, accelerating derivation of COL7A1-edited iPS
Monica Cassandras et al.
Nature cell biology, 22(11), 1295-1306 (2020-10-14)
Aberrant epithelial reprogramming can induce metaplastic differentiation at sites of tissue injury that culminates in transformed barriers composed of scar and metaplastic epithelium. While the plasticity of epithelial stem cells is well characterized, the identity and role of the niche
Jaymin J Kathiriya et al.
Nature cell biology, 24(1), 10-23 (2022-01-01)
Loss of alveolar type 2 cells (AEC2s) and the ectopic appearance of basal cells in the alveoli characterize severe lung injuries such as idiopathic pulmonary fibrosis (IPF). Here we demonstrate that human alveolar type 2 cells (hAEC2s), unlike murine AEC2s

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service