Skip to Content
MilliporeSigma

CBL173

Anti-Desmoplakin 1& 2 Antibody, clone DP2.15

clone DP2.15, Chemicon®, from mouse

Synonym(s):

Anti-DCWHKTA, Anti-DP

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
DP2.15, monoclonal
Application:
ICC, IF, WB
Citations:
6
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

DP2.15, monoclonal

species reactivity

bovine, human, chicken, rat, mouse

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable, immunofluorescence: suitable, western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... DSP(1832)

Immunogen

Bovine desmoplakin 1 & 2

Application

Anti-Desmoplakin 1& 2 Antibody, clone DP2.15 detects level of Desmoplakin 1& 2 & has been published & validated for use in IF, WB & IC.
Research Category
Cell Structure
Research Sub Category
Cytoskeleton
Western blot
Immunohistochemistry: 1:10; Frozen tissue
Immunofluorescence

Optimal working dilutions must be determined by the end user.

Biochem/physiol Actions

This antibody shows distinct punctate membrane staining of desmoplakins 1 (MW 250 kDa) and 2 (MW 215 kDa) in stratified epithelia and simple epithelia, including glands, urothelium, thymic reticular epithelium, hepatocytes, intercalated disks of myocardium and arachnoid cells of meninges.

Physical form

Format: Purified
Lyophilized. Reconstitute in 1 mL distilled water (final solution contains 0.09% sodium
azide, 0.5% BSA in PBS buffer, pH 7.4

Preparation Note

Reconstituted antibody can be stored at 2°-8°C for up to 12 months from date of receipt. Antibody can also be stored at -20°C in undiluted aliquots.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Andrew Muroyama et al.
eLife, 6 (2017-09-05)
The physiological functions of microtubules (MTs) are poorly understood in many differentiated cell types. We developed a genetic toolkit to study MT dynamics and function in diverse cells. Using TRE-EB1-GFP mice, we found that MT dynamics are strongly suppressed in
Annika Gross et al.
Cellular and molecular gastroenterology and hepatology, 13(4), 1181-1200 (2021-12-21)
Desmosomes are intercellular junctions connecting keratin intermediate filaments of neighboring cells. The cadherins desmoglein 2 (Dsg2) and desmocollin 2 mediate cell-cell adhesion, whereas desmoplakin (Dsp) provides the attachment of desmosomes to keratins. Although the importance of the desmosome-keratin network is
Emily I Bartle et al.
The Journal of cell biology, 219(6) (2020-05-14)
Desmosomes are cell-cell junctions that provide mechanical integrity to epithelial and cardiac tissues. Desmosomes have two distinct adhesive states, calcium-dependent and hyperadhesive, which balance tissue plasticity and strength. A highly ordered array of cadherins in the adhesive interface is hypothesized



Global Trade Item Number

SKUGTIN
CBL17304053252468490