Skip to Content
MilliporeSigma
All Photos(4)

Documents

CANPIT-96K

Millipore

MILLIPLEX® Canine Pituitary Expanded Panel - Endocrine Multiplex Assay

The analytes available for this multiplex kit are: ACTH, FSH, GH, TSH, LH, & Prolactin.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.47

species reactivity

canine

manufacturer/tradename

Milliplex®

assay range

accuracy: 92%
(TSH)

accuracy: 95%
(GH)

sensitivity: 0.01 ng/mL
(GH)

sensitivity: 0.09 ng/mL
(LH)

sensitivity: 1.21 ng/mL
(FSH)

sensitivity: 1.62 ng/mL
(TSH)

sensitivity: 2.45 ng/mL
(Prolactin)

sensitivity: 2.45 pg/mL
(ACTH)

standard curve range: 0.01-50 ng/mL
(GH)

standard curve range: 0.1-350 ng/mL
(LH)

standard curve range: 0.9-3,750 ng/mL
(TSH)

standard curve range: 1.7-7,000 ng/mL
(Prolactin)

standard curve range: 2.4-10,000 ng/mL
(FSH)

standard curve range: 2.4-10,000 pg/mL
(ACTH)

inter-assay cv: <20%
intra-assay cv: <10%

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex xMAP)

shipped in

wet ice

storage temp.

2-8°C

General description

Called the “master gland” because it controls many other endocrine glands, the pituitary gland secretes key hormones that play important roles in the regulation of metabolism, growth, and reproduction. The Canine Pituitary Expanded Panel thus enables you to focus on the therapeutic potential of reproduction, growth, metabolic homeostasis, and pituitary related diseases such as acromegaly, growth hormone deficiency, diabetes insipidus and pituitary tumors.

MILLIPLEX® Canine Pituitary Expanded Panel is a 6-plex kit to be used for the quantification of any or all of the following analytes: Adrenocorticotropic Hormone (ACTH), Growth Hormone (GH), Follicle-Stimulating Hormone (FSH), Luteinizing Hormone (LH), Prolactin, and Thyroid-Stimulating Hormone (TSH). This kit may be used for the analysis of pituitary hormones in canine serum or plasma, canine tissue extract, or cell/tissue supernatant samples. This kit uses a 96-well format, contains a lyophilized standard cocktail, two internal assay quality controls and can measure up to 38 samples in duplicate.

The Luminex® xMAP® platform uses a magnetic bead immunoassay format for ideal speed and sensitivity to quantitate multiple analytes simultaneously, dramatically improving productivity while conserving valuable sample volume.

Panel Type: Endocrine

Specificity

Cross Reactivty
There was no or negligible cross-reactivity between the antibodies for an analyte and any of the other analytes in this panel except for canine LH having ~5% cross-reactivity with the TSH antibody pair.

Application

  • Analytes: Adrenocorticotropic Hormone (ACTH), Growth Hormone (GH), Follicle-Stimulating Hormone (FSH), Luteinizing Hormone (LH), Prolactin, and Thyroid-Stimulating Hormone (TSH)
  • Recommended Sample Type: Canine serum or plasma, canine tissue extract, or cell/tissue supernatants
  • Recommended Sample Dilution: 25 μL per well of undiluted serum or plasma
  • Assay Run Time: Overnight (16-18 hours) at 2-8°C
  • Research Category: Endocrine

Features and Benefits

Design your multiplex kit by choosing available analytes within this panel.

Linkage

Replaces: This panel replaces CPTMAG-96K.

Storage and Stability

Recommended storage for kit components is 2 - 8°C.

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2

Target Organs

Respiratory Tract

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Discover benefits of using biomarker detection multiplex immunoassays for companion and agricultural animal health research and find vet med multiplex assays.

Discover benefits of using biomarker detection multiplex immunoassays for companion and agricultural animal health research and find vet med multiplex assays.

Discover benefits of using biomarker detection multiplex immunoassays for companion and agricultural animal health research and find vet med multiplex assays.

Discover benefits of using biomarker detection multiplex immunoassays for companion and agricultural animal health research and find vet med multiplex assays.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service