MilliporeSigma
BRZMCC
Mobius® Breez Microbioreactor Media Characterization Consumable for Media Calibration
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
12352200
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
description
Mobius®
technique(s)
cell culture | mammalian: suitable
Features and Benefits
A 2 mL Automated Perfusion Cell Culture Platform
For perfusion cell line development, media screening and optimization, and early process development, the ability to run multiple experiments at rapid speed is essential. The Mobius® Breez Microbioreactor System is designed for rapid cell culture experiments, providing an easy-to use solution to produce high quality datasets. It allows for increased productivity and shorter development timelines.
Other Notes
Mobius® Breez Microbioreactor System Services
The pharmaceutical and biotechnology industries are highly regulated. To support you as you navigate this challenging environment, we offer a wide range of services that can help save time and lower costs. All of our services are performed by global experts who have an intimate knowledge of our equipment backed by decades of experience, giving you peace of mind at every step.
Legal Information
MOBIUS is a registered trademark of Merck KGaA, Darmstadt, Germany