Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AX0115

Supelco

Acetone

HPLC, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs

Synonym(s):

Dimethyl Ketone, 2-Propanone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH3
CAS Number:
Molecular Weight:
58.08
Beilstein:
635680
MDL number:
UNSPSC Code:
12352115
EC Index Number:
200-662-2
grade:
reagent
technique(s):
HPLC: suitable
bp:
56 °C/760 mmHg (lit.)
vapor pressure:
184 mmHg ( 20 °C)

grade

reagent

Quality Level

Agency

meets USP/NF

vapor density

2 (vs air)

vapor pressure

184 mmHg ( 20 °C)

Assay

≥99.5% (corrected for water content, GC)

form

liquid

expl. lim.

13.2 %

technique(s)

HPLC: suitable

impurities

≤0.0003 meq/g Titrable acid
≤0.0006 meq/g Titrable base
≤0.002% Aledhyde (as HCHO)
≤0.05% Isopropyl Alcohol
≤0.05% Methanol
≤0.5% Water

evapn. residue

≤5 ppm

color

clearAPHA: ≤10

refractive index

n20/D 1.359 (lit.)

pH

5-6 (20 °C, 395 g/L in H2O)

bp

56 °C/760 mmHg (lit.)

mp

−94 °C (lit.)

transition temp

flash point <-20 °C

density

0.791 g/mL at 25 °C (lit.)

λmax

≤330 nm

UV absorption

λ: 330 nm Amax: ≤1.00
λ: 340 nm Amax: ≤0.10
λ: 350 nm Amax: ≤0.02
λ: 400 nm Amax: ≤0.01

format

neat

shipped in

ambient

storage temp.

room temp

SMILES string

CC(C)=O

InChI

1S/C3H6O/c1-3(2)4/h1-2H3

InChI key

CSCPPACGZOOCGX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

AX0115P-1 and AX0115P-6 are packaged in poly-coated glass bottles.
Acetone′s luminesence intensity is dependent upon the solution components . The absorption of UV light by acetone, results in its photolysis and the production of radials .
HPLC, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs

Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

Aldehyde (as HCHO): 0.002% max
Appearance: Clear liquid, free from particulates
Assay (GC, corrected for water content): 99.5% min
Color (APHA): 10 max
Evaporation residue: 5ppm max
Filtered through 0.2 m filter: To pass test
Form: Liquid
Infrared Spectrum: Conforms to standard
Isopropyl alcohol: 0.05% max
Methanol: 0.05% max
Solubility in water: To pass test
Substances reducing permanganate: To pass test
Titrable acid: 0.0003meq/g max
Titrable base: 0.0006meq/g max
UV Abs. at 330 nm: 1.00AU max
UV Abs. at 340 nm: 0.10AU max
UV Abs. at 350 nm: 0.02AU max
UV Abs. at 400 nm: 0.01AU max
UV Cut-off: 330nm max
Water (H2O): 0.5% max

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service