Skip to Content
MilliporeSigma
All Photos(1)

Documents

AX0077

Supelco

Acetic acid (glacial)

≥99%, OmniTrace

Synonym(s):

Acetic acid, Ethanoic Acid, Glacial acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2H
CAS Number:
Molecular Weight:
60.05
Beilstein:
506007
MDL number:
UNSPSC Code:
12352106
EC Index Number:
200-580-7

product name

Acetic Acid, Glacial 99%, OmniTrace, ≥99%

grade

for inorganic trace analysis

Quality Level

vapor density

2.07 (vs air)

vapor pressure

15.4 hPa ( 20 °C)

Assay

≥99%

form

liquid

autoignition temp.

800 °F

expl. lim.

16 %, 92 °F
4 %, 59 °F

concentration

≤100% in acetic acid

refractive index

n20/D 1.371 (lit.)

pH

2.5 (20 °C, 50 g/L in H2O)

bp

117-118 °C (lit.)

mp

16.2 °C (lit.)

transition temp

flash point 39 °C

solubility

602.9 g/L (at 25 °C (77 °F) at 1,013 hPa - completely soluble in water)

density

1.04 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤1 ppm
phosphate (PO43-): ≤1 ppm
sulfate (SO42-): ≤0.5 ppm

cation traces

Ag: ≤1 ppb
Al: ≤1 ppb
As: ≤0.5 ppb
Ba: ≤0.5 ppb
Be: ≤0.1 ppb
Bi: ≤0.1 ppb
Ca: ≤1 ppb
Cd: ≤0.5 ppb
Ce: ≤0.1 ppb
Co: ≤0.1 ppb
Cr: ≤1 ppb
Cs: ≤0.1 ppb
Cu: ≤0.5 ppb
Dy: ≤0.1 ppb
Er: ≤0.1 ppb
Eu: ≤0.1 ppb
Fe: ≤1 ppb
Ga: ≤0.1 ppb
Gd: ≤0.1 ppb
Ge: ≤0.5 ppb
Hf: ≤0.1 ppb
Hg: ≤1 ppb
Ho: ≤0.1 ppb
In: ≤0.1 ppb
K: ≤1 ppb
La: ≤0.1 ppb
Li: ≤0.1 ppb
Lu: ≤0.1 ppb
Mg: ≤0.5 ppb
Mn: ≤0.5 ppb
Mo: ≤0.5 ppb
Na: ≤1 ppb
Nd: ≤0.1 ppb
Ni: ≤0.5 ppb
Pb: ≤0.1 ppb
Pr: ≤0.1 ppb
Pt: ≤0.5 ppb
Rb: ≤0.1 ppb
Re: ≤0.1 ppb
Rh: ≤0.5 ppb
Ru: ≤0.5 ppb
Sb: ≤0.5 ppb
Sc: ≤0.1 ppb
Se: ≤1 ppb
Sm: ≤0.1 ppb
Sn: ≤0.5 ppb
Sr: ≤0.5 ppb
Tb: ≤0.1 ppb
Te: ≤0.5 ppb
Th: ≤0.1 ppb
Ti: ≤0.5 ppb
Tl: ≤0.1 ppb
Tm: ≤0.1 ppb
U: ≤0.1 ppb
V: ≤0.5 ppb
W: ≤0.5 ppb
Y: ≤0.1 ppb
Yb: ≤0.1 ppb
Zn: ≤1 ppb
Zr: ≤0.1 ppb

SMILES string

[F2C(F2C)13F3C]C(O)=O

shipped in

ambient

storage temp.

room temp

InChI

1S/C2H4O2/c1-2(3)4/h1H3,(H,3,4)

InChI key

QTBSBXVTEAMEQO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

OmniTrace acids offer an economical alternative for sensitive, trace metal analysis and sample preparation. These acids are tested to PPB (parts per billion) levels.

Packaging

Product is packed into HDPE bottles. Each HDPE bottle is packed into a single bottle shipping case.

Legal Information

OMNITRACE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

102.2 °F - closed cup

Flash Point(C)

39 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service