Skip to Content
MilliporeSigma

AR001

Freund′s Complete Adjuvant

Synonym(s):

FCA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41116012
NACRES:
NA.77
eCl@ss:
32160405
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid

Quality Segment

manufacturer/tradename

Chemicon®

shipped in

wet ice

Application

Composition:

85% Drakeol 6VR (Mineral Oil)

15% Mannide Monooleate (Arlacel A)

20 mg Mycobacterium
Research Category
All

Preparation Note

Store at 2-8°C for up to twelve months.

Note: This material is of U.S. origin and donor animals were free from visible disease at the time of blood collection.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Storage Class

10 - Combustible liquids

WGK

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library