Skip to Content
MilliporeSigma
All Photos(1)

Documents

APT139

Sigma-Aldrich

Caspase 9 Colorimetric Activity Assay Kit, LEHD

Caspase-9 Colorimetric Activity Assay Kits provide a simple & convenient means for assaying the activity of caspases that recognize the LEHD.

Synonym(s):

Mch-6, ICE-LAP6, Apaf-3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

Quality Level

species reactivity (predicted by homology)

mammals

manufacturer/tradename

Chemicon®

technique(s)

activity assay: suitable

NCBI accession no.

UniProt accession no.

detection method

colorimetric

shipped in

dry ice

Gene Information

human ... CASP9(842)

General description

Activation of ICE-family proteases/caspases initiates apoptosis in mammalian cells. Chemicon′s Caspase-9 Colorimetric Activity Assay Kits provide a simple and convenient means for assaying the activity of caspases that recognize the LEHD. The assay is based on spectophotometric detection of the chromophore p-nitroaniline (pNA) after cleavage from the labeled substrate LEHD-pNA. The free pNA can be quantified using a spectrophotometer or a microtiter plate reader at 405 nm. Comparison of the absorbance of pNA from an apoptotic sample with an uninduced control allows determination of the fold increase in caspase-9 activity.

Application

Caspase-9 Colorimetric Activity Assay Kits provide a simple & convenient means for assaying the activity of caspases that recognize the LEHD.
Research Category
Apoptosis & Cancer

Components

5X Cell Lysis Buffer (Part No. 90065): 5 mL

5X Assay Buffer (Part No. 90066): 10 mL

Caspase-9 Substrate (Ac-LEHD-pNA) (Part No. 90083): 250 μL of 3 mg/mL solution

Caspase-9 Inhibitor (Ac-LEHD-CHO) (Part No. 90087): 50 μL of 100 μM (0.054 mg/mL) in DMSO

pNA Standard (Part No. 90085): 250 μL of 10 mM in DMSO

Storage and Stability

Store kit materials at -20°C up to their expiration date.

Special Precautions:

· After thawing reagents, use immediately or aliquot and freeze at -20°C for longer storage. Avoid repeated freeze/thaw cycles.

· The Caspase-9 substrate and pNA standard are especially light sensitive. Maintain these reagents in amber or covered containers.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vanessa Blas-Valdivia et al.
Frontiers in pharmacology, 12, 671614-671614 (2021-07-24)
Thyroid hormone is essential for hippocampal redox environment and neuronal viability in adulthood, where its deficiency causes hypothyroidism related to oxidative and endoplasmic reticulum stresses in the hippocampus, resulting in neuronal death. One option of treatment is antioxidants; however, they

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service