Skip to Content
MilliporeSigma

AP114

Goat Anti-Human IgM Antibody, Fc5µ

2 mg/mL, Chemicon®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ELISA, IP, WB
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

goat

Quality Segment

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Chemicon®

concentration

2 mg/mL

technique(s)

ELISA: suitable, immunoprecipitation (IP): suitable, western blot: suitable

shipped in

wet ice

target post-translational modification

unmodified

Immunogen

Highly purified Human IgM isolated from pooled normal Human serum.

Application

Optimal working dilutions must be determined by end user.
Research Category
Secondary & Control Antibodies
Research Sub Category
Fragment Specific Secondary Antibodies
This Goat anti-Human IgM Antibody, Fc5μ is validated for use in ELISA, IP, WB for the detection of Human IgM.

Biochem/physiol Actions

Based on immunoelectrophoresis, the antibody reacts with the Fc5μ portion of the human IgM heavy chain. No antibody was detected against normal human IgG or IgA, or against non-immunoglobulin serum proteins, but the antibody may cross-react with immunoglobulins from other species. The purified antibody shows a single band on IEP at a concentration of 10 mg/mL when run against anti-goat whole serum.

Physical form

Affinity purified antibody presented as a liquid in 0.02 M Phosphate Buffer, 0.25 M NaCl containing 0.1% sodium azide. Filtered through a 0.45 micron membrane filter.

Preparation Note

Store between 2 to 8°C for up 6 months donot freeze thaw; use sterile technique to maintain quality.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


wgk

WGK 2

Storage Class

12 - Non Combustible Liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library