Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
12352203
NACRES:
NA.26
eCl@ss:
32160702
Form:
liquid
Assay:
>90% (SDS-PAGE)
Biological source:
human
biological source
human
Quality Segment
assay
>90% (SDS-PAGE)
form
liquid
manufacturer/tradename
Chemicon®
technique(s)
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
dry ice
Gene Information
human ... PPP1R1B(84152)
General description
Product Source: Recombinant human DARPP-32, expressed in bacteria with a His-Tag linker.
Application
Phosphatase assays
Research Category
Neuroscience
Neuroscience
Research Sub Category
Reward & Addiction
Signaling Neuroscience
Reward & Addiction
Signaling Neuroscience
Physical form
0.5 mg/mL in Tris-buffered saline containing 40% glycerol.
Preparation Note
Store at -20°C for up to 1 year.
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class
10 - Combustible liquids
wgk
WGK 1
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.