Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
12352203
NACRES:
NA.46
eCl@ss:
32160702
Conjugate:
biotin conjugate
Clone:
polyclonal
Application:
IHC
Citations:
-
biological source
sheep
Quality Segment
conjugate
biotin conjugate
antibody form
purified antibody
antibody product type
secondary antibodies
clone
polyclonal
species reactivity
mouse
technique(s)
immunohistochemistry: suitable
isotype
IgG1
shipped in
wet ice
target post-translational modification
unmodified
General description
Immunoglobulin G (IgG) is part of the family of glycoprotein immunoglobulins which stage immune responses against pathogenic organisms. IgG contributes to the immune response by mobilizing innate immune effector cells and inducing and pro-inflammatory responses. Such proinflammatory responses are mediated by Fcg receptors which interact with sialylated Fc fragments on IgG molecules.
~150 kDa observed.
Immunogen
Purified mouse IgG.
Application
Immunohistochemistry Analysis: A representative lot was validated by immunohistochemistry staining (1:500 dilution) of human liver tissue section following a mouse monoclonal anti-CREB primary antibody (Cat. No. MAB5432-C).
Research Category
Secondary & Control Antibodies
Secondary & Control Antibodies
Research Sub Category
Fragment Specific Secondary Antibodies
Fragment Specific Secondary Antibodies
Sheep Anti-Mouse IgG Antibody, Species absorbed, Biotin conjugate is an antibody against Sheep Mouse IgG for use in Immunohistochemistry.
Biochem/physiol Actions
Based on ELISA analysis using the unconjugated antibody (Cat. No. AC111), this antibody reacts with whole molecule mouse IgG (IgG>>IgG2a>IgG>>>>IgG1) with minimal or no cross-reactivity toward mouse IgA or IgM.
This antibody has been species absorbed and tested by ELISA prior to conjugation to ensure minimal cross-reactivity toward chicken IgY or IgG from human, rat, rabbit, guinea pig, and hamster species, but may cross-react with immunoglobulins from other species.
Physical form
Fractionated by octanoic acid and ammonium sulfate cut, and then depleted with rat IgG column and mouse IgA column prior to biotin conjugation.
Purified sheep polyclonal antibody in PBS with 15 mg/mL BSA and 0.1 % sodium azide.
Preparation Note
Stable for 1 year at 2-8°C from date of receipt.
Analysis Note
Evaluated by Immunohistochemistry in human prostate tissue.
Immunohistochemistry Analysis: Performance of this biotin-conjugated secondary antibody has been validated by immunohistochemistry staining (1:500 dilution) of human prostate tissue section following a mouse monoclonal anti-CREB primary antibody (Cat. No. MAB5432-C).
Immunohistochemistry Analysis: Performance of this biotin-conjugated secondary antibody has been validated by immunohistochemistry staining (1:500 dilution) of human prostate tissue section following a mouse monoclonal anti-CREB primary antibody (Cat. No. MAB5432-C).
Other Notes
Concentration: Please refer to lot specific datasheet.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.