Skip to Content

ABT95

Anti-TGN46, Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Trans-Golgi network integral membrane protein 2, TGN38 homolog, TGN46, TGN48, Trans-Golgi network protein TGN51

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ICC, IP, WB
Citations:
4
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, rat, human

species reactivity (predicted by homology)

canine (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TGOLN2(10618)

General description

TGN46 is one of several proteins that utilize retrograde transport to travel between the trans-Golgi network (TGN) and the endocytic pathway. Retrograde transport involves the tethering, docking, fusion, and post translational modification of TGN proteins. Further studies are still required to elucidate the roles and mode of action for TGN46.
~110 kDa observed. This protein is observed at ~110 kDa, and it is subject to glycosylation (Khanobdee, K. et al. (2004). Mol Vis. 13(10):933-42.; Prescott, A. R., et al. (1997). Eur J Cell Biol. 72(3):238-246.).

Immunogen

Epitope: Cytoplasmic domain
KLH-conjugated linear peptide corresponding to the cytoplasmic domain of human TGN46.

Application

Anti-TGN46 detects levels of TGN46 proteins & has been published & validated for use in wb, IP & ICC.
Research Category
Cell Structure
Research Sub Category
Vesicular Trafficking
Western Blot Analysis: 2 µg/mL of this antibody detected TGN46 in 10 µg of NIH/3T3 cell lysate. Multiple uncharacterized bands may be observed in some cell lysates.

Immunocytochemistry Analysis: A 1:500 dilution from a representative lot detected TGN46 in A431 and HeLa cells.

Biochem/physiol Actions

This antibody recognizes the cytoplasmic domain of TGN46.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa cell lysate
Evaluated by Immunoprecipitation in HeLa cell lysate.

Immunoprecipitation Analysis: 0.2 µg of this antibody immunoprecipitated TGN46 in 500 µg of HeLa cell lysate (An uncharacterized band at ~29 kDa may be observed in some cell lysates.).

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library