Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABS994

Sigma-Aldrich

Anti-phospho-Glucocorticoid receptor Antibody, Ser226

from rabbit, purified by affinity chromatography

Synonym(s):

Nuclear receptor subfamily 3 group C member 1, glucocorticoid receptor, GR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

mouse (based on 100% sequence homology), rat (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer226)

Gene Information

human ... NR3C1(2908)

General description

Glucocorticoid receptor (GR), also known as Nuclear receptor subfamily 3 group C member 1, and encoded by the gene name NR3C1/ GRL, is receptor for glucocorticoids (GC). GR has a dual mode of action: as a transcription factor that binds to glucocorticoid response elements (GRE), both for nuclear and mitochondrial DNA, and as a modulator of other transcription factors. GR affects inflammatory responses, cellular proliferation and differentiation in target tissues. Recent studies have shown that GR may act as a coactivator for STAT5-dependent transcription upon growth hormone (GH) stimulation and could reveal an essential role of hepatic GR in the control of body growth. In addition, GR is involved in chromatin remodeling, and may play a negative role in adipogenesis through the regulation of lipolytic and antilipogenic genes expression. Mutation in GR has been associated with Glucocorticoid resistance (GCRES): a hypertensive, hyperandrogenic disorder characterized by increased serum cortisol concentrations. Inheritance is autosomal dominant. GR is widely expressed. In the heart, GR is detected in left and right atria, left and right ventricles, aorta, apex, intraventricular septum, and atrioventricular node, as well as whole adult and fetal heart.

Immunogen

Epitope: A region within the modulating domain
KLH-conjugated linear peptide corresponding to the domain of: Modulating region of Human Glucocorticoid receptor.

Application

Research Category
Signaling
Research Sub Category
Lipid Metabolism & Weight Regulation
This Anti-phospho-Glucocorticoid receptor antibody is validated for use in Western Blotting for the detection of phospho-Glucocorticoid receptor.
Western Blotting Analysis: A representative lot of this antibody at 0.5 µg/mL dilution detected phosphorylated glucocorticoid receptor protein in transfected U2OS cell lysate (Chen, W., et al. Mol Endocrinol, August 2008, 22(8):1754–1766)
Western Blotting Analysis: A representative lot of this antibody at 0.5 µg/mL dilution detected phosphorylated glucocorticoid receptor protein in rat hepatoma cell lysate (Blind,R., et al. Journal of Steroid Biochemistry & Molecular Biology 109 (2008) 150–157)

Quality

Evaluated by Western Blotting on transfected HEK293 cell lysate

Western Blotting Analysis: 1 µg/mL dilution of this antibody detected phosphorylated glucocorticoid receptor in 6 µg of transfected HEK293 cell lysate

Target description

~85 kDa observed

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service