Skip to Content
MilliporeSigma

ABS97

Anti-KEAP1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Cytosolic inhibitor of Nrf2, Kelch-like protein 19, INrf2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ICC, WB
Citations:
3
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human, rat

technique(s)

immunocytochemistry: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... KEAP1(9817)

General description

Anti-oxidant defense is known to be regulated by various mechanisms including the usage of nuclear factor erythroid 2-related factor 2 (Nrf2). Under normal physiological conditions, Nrf2 is bound to Kelch-like ECH-associated protein 1 (Keap1), a cytoplasmic repressor. Keap1 also acts as a substrate adaptor for Cullin3-dependent ubiquitin ligase and also targets Nrf2 for degradation by proteosomes. The substrate adaptor function of Keap1 is inactivated during physiological stress. In this respective condition, a range of oxidative and electrophilic stimuli such as ROS, diethyl malonate, and certain disease processes, resulting from Nrf2 accumulation, enters the nucleus and thereby activates the expression of select anti-oxidant genes. Such genes promote the detoxification of ROS and other harmful molecules, which contribute to the carcinogenesis.
~60-70 kDa observed.
A doublet may be observed in some cell lysates.

Immunogen

Epitope: Kelch 3 Region
KLH-conjugated linear peptide corresponding to the Kelch 3 region of human KEAP1.

Application

Anti-KEAP1 Antibody detects level of KEAP1 & has been published & validated for use in WB & IC.
Immunocytochemistry Analysis: 2 µg/mL from a representative lot detected KEAP1 in A431 and HeLa cells.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
General Post-translation Modification

Biochem/physiol Actions

This antibody recognizes the Kelch 3 region of KEAP1.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa cell lysate
Evaluated by Western Blot in HeLa cell lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected KEAP1 in 10 µg of HeLa cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library